Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033107.3 | NP_149098.2 | 3 - 4 | 390 | 91 |
GenBank mRNA | AK095561.1 | - | 3 - 4 | 380 | 91 |
AL525056.3 | - | 3 - 4 | 332 | 91 | |
BC004923.2 | - | 2 - 3 | 287 | 91 | |
BC021573.1 | - | 2 - 3 | 287 | 91 | |
BC107714.1 | - | 3 - 4 | 374 | 91 |