Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102566.1 | NP_001096036.1 | 1 - 2 | 260 | 144 |
GenBank mRNA | BC028905.2 | - | 1 - 2 | 260 | 144 |
BC067884.1 | - | 1 - 2 | 238 | 142 | |
BC147003.1 | - | 1 - 2 | 161 | 144 | |
BC147004.1 | - | 1 - 2 | 161 | 144 | |
CN283207.1 | - | 1 - 2 | 226 | 144 |