Assay Design



Gene Transcripts

Gene Symbol
TMEM26
Entrez Gene ID
219623
Gene Name
transmembrane protein 26
Gene Aliases
-
Chromosome Location
Chr.10: 61406642 - 61453450 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178505.7 NP_848600.2 1 - 2 554 109
NR_134507.1 1 - 2 554 109
NR_134508.1 1 - 2 554 109
XM_011539450.2 XP_011537752.1 1 - 2 342 109
XM_011539451.1 XP_011537753.1 1 - 2 342 109
XM_017015829.1 XP_016871318.1 1 - 2 342 109
GenBank mRNA AK125611.1 - 1 - 2 342 109
BC042872.1 - 1 - 2 271 109
CR749606.1 - 1 - 2 554 109

View Full Product Details