Assay Design



Gene Transcripts

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
-
Chromosome Location
Chr.1: 92080305 - 92147840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183242.3 NP_899065.2 8 - 9 1287 77
GenBank mRNA AK296516.1 - 7 - 8 957 77
BC013922.1 - 8 - 9 1260 77

View Full Product Details