Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098844.2 | NP_001092314.1 | 2 - 3 | 358 | 62 |
XM_011519626.2 | XP_011517928.1 | 2 - 3 | 509 | 62 | |
XM_011519627.2 | XP_011517929.1 | 2 - 3 | 235 | 62 | |
XM_017016574.1 | XP_016872063.1 | 2 - 3 | 1084 | 62 | |
GenBank mRNA | AK300456.1 | - | 1 - 2 | 101 | 62 |
BC157856.1 | - | 2 - 3 | 358 | 62 |