Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172774.1 | NP_001166245.1 | 8 - 9 | 1054 | 83 |
NM_207325.2 | NP_997208.2 | 8 - 9 | 1080 | 83 | |
XM_006723045.1 | XP_006723108.1 | 5 - 6 | 596 | 83 | |
XM_011526526.2 | XP_011524828.1 | 8 - 9 | 1035 | 83 | |
XM_017026366.1 | XP_016881855.1 | 8 - 9 | 1071 | 83 | |
XM_017026367.1 | XP_016881856.1 | 8 - 9 | 994 | 83 | |
GenBank mRNA | AK126264.1 | - | 2 - 3 | 155 | 83 |
AK126757.1 | - | 8 - 9 | 1071 | 83 | |
AK129497.1 | - | 8 - 9 | 1035 | 83 | |
BC131725.1 | - | 7 - 8 | 902 | 83 | |
CR749459.1 | - | 2 - 3 | 250 | 83 |