Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 1 - 2 | 176 | 131 |
XM_011534936.1 | XP_011533238.1 | 1 - 2 | 165 | 131 | |
XM_017020395.1 | XP_016875884.1 | 1 - 2 | 176 | 131 | |
GenBank mRNA | AK291273.1 | - | 1 - 2 | 165 | 131 |
BC068595.1 | - | 1 - 2 | 156 | 131 | |
DN999521.1 | - | 1 - 2 | 216 | 131 |