Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027410.1 | 4 - 5 | 552 | 77 | |
XM_011521596.1 | XP_011519898.1 | 3 - 4 | 544 | 77 | |
XM_011521597.1 | XP_011519899.1 | 3 - 4 | 544 | 77 | |
XM_017022210.1 | XP_016877699.1 | 3 - 4 | 544 | 77 | |
XM_017022211.1 | XP_016877700.1 | 4 - 5 | 617 | 77 | |
XM_017022213.1 | XP_016877702.1 | 3 - 4 | 544 | 77 | |
XM_017022214.1 | XP_016877703.1 | 3 - 4 | 544 | 77 | |
XM_017022215.1 | XP_016877704.1 | 3 - 4 | 544 | 77 | |
XM_017022216.1 | XP_016877705.1 | 3 - 4 | 544 | 77 | |
XM_017022217.1 | XP_016877706.1 | 3 - 4 | 544 | 77 | |
GenBank mRNA | BX647673.1 | - | 3 - 4 | 521 | 77 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027409.1 | 4 - 5 | 566 | 77 | |
XM_017022018.1 | XP_016877507.1 | 2 - 3 | 347 | 77 | |
XM_017022019.1 | XP_016877508.1 | 2 - 3 | 347 | 77 | |
GenBank mRNA | BX648160.1 | - | 3 - 4 | 521 | 77 |
BX648575.1 | - | 2 - 3 | 360 | 77 |