Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138369.2 | NP_612378.1 | 2 - 3 | 593 | 105 |
GenBank mRNA | AK301982.1 | - | 2 - 3 | 581 | 105 |
AY303777.1 | - | 2 - 3 | 366 | 105 | |
BC007436.2 | - | 2 - 3 | 583 | 105 |