Assay Design



Gene Transcripts

Gene Symbol
C16orf87
Entrez Gene ID
388272
Gene Name
chromosome 16 open reading frame 87
Gene Aliases
-
Chromosome Location
Chr.16: 46802047 - 46831162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001436.2 NP_001001436.1 1 - 2 78 128
GenBank mRNA BC056676.1 - 1 - 2 82 128
BX647945.1 - 1 - 2 78 128

View Full Product Details