Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201628.2 | NP_963922.2 | 13 - 14 | 2327 | 120 |
XM_005245795.4 | XP_005245852.1 | 15 - 16 | 2582 | 120 | |
XM_011541074.2 | XP_011539376.1 | 15 - 16 | 2589 | 120 | |
XM_011541075.2 | XP_011539377.1 | 13 - 14 | 2245 | 120 | |
XM_011541076.2 | XP_011539378.1 | 13 - 14 | 2390 | 120 | |
XM_011541077.2 | XP_011539379.1 | 13 - 14 | 3369 | 120 | |
XM_011541081.2 | XP_011539383.1 | 7 - 8 | 1069 | 120 | |
GenBank mRNA | AK293806.1 | - | 8 - 9 | 1133 | 120 |
BC101637.1 | - | 3 - 4 | 408 | 120 | |
BC113621.1 | - | 3 - 4 | 408 | 120 | |
BC143641.1 | - | 3 - 4 | 354 | 120 | |
DA317442.1 | - | 3 - 4 | 345 | 120 | |
DN831635.1 | - | 4 - 5 | 609 | 120 |