Assay Design



Gene Transcripts

Gene Symbol
TCF24
Entrez Gene ID
100129654
Gene Name
transcription factor 24
Gene Aliases
TCF-24, bHLHa25
Chromosome Location
Chr.8: 66946501 - 66962592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193502.1 NP_001180431.1 1 - 2 263 148
GenBank mRNA AW028708.1 - 2 - 3 148

View Full Product Details