Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017961.4 | NP_001017961.1 | 1 - 2 | 772 | 74 |
NR_135199.1 | 1 - 2 | 772 | 74 | ||
XM_017000417.1 | XP_016855906.1 | 1 - 2 | 772 | 74 | |
XM_017000418.1 | XP_016855907.1 | 1 - 2 | 323 | 74 | |
GenBank mRNA | AK299946.1 | - | 1 - 2 | 302 | 74 |
BC114214.1 | - | 2 - 3 | 855 | 74 |