Assay Design
Gene Transcripts
- Gene Symbol
- TRABD2A
- Entrez Gene ID
-
129293
- Gene Name
- TraB domain containing 2A
- Gene Aliases
-
C2orf89, TIKI1
- Chromosome Location
-
Chr.2: 84821650 - 84881975 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC073808.1 |
- |
2 - 3 |
290 |
77 |
View Full Product Details