Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272101.1 | NP_001259030.1 | 3 - 4 | 458 | 79 |
NM_002429.5 | NP_002420.1 | 3 - 4 | 458 | 79 | |
NR_073606.1 | 3 - 4 | 429 | 79 | ||
XM_011538359.1 | XP_011536661.1 | 3 - 4 | 444 | 79 | |
XM_017019308.1 | XP_016874797.1 | 3 - 4 | 444 | 79 | |
XM_017019309.1 | XP_016874798.1 | 3 - 4 | 444 | 79 | |
GenBank mRNA | AK225939.1 | - | 3 - 4 | 448 | 79 |
AK303202.1 | - | 3 - 4 | 411 | 79 | |
AL545199.3 | - | 3 - 4 | 458 | 79 | |
BC050368.1 | - | 3 - 4 | 397 | 79 | |
U37791.1 | - | 3 - 4 | 386 | 79 | |
U38321.1 | - | 3 - 4 | 381 | 79 | |
U38431.1 | - | 3 - 4 | 350 | 79 | |
X92521.1 | - | 3 - 4 | 406 | 79 | |
Y08622.1 | - | 3 - 4 | 405 | 79 |