Assay Design



Gene Transcripts

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
Chromosome Location
Chr.1: 74026015 - 74198201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322315.1 NP_001309244.1 7 - 8 1428 114
GenBank mRNA AK131476.2 - 9 - 10 3750 114

View Full Product Details