Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308154.1 | NP_001295083.1 | 5 - 6 | 494 | 75 |
NM_198686.2 | NP_941959.1 | 5 - 6 | 625 | 75 | |
XM_017021277.1 | XP_016876766.1 | 6 - 7 | 754 | 75 | |
GenBank mRNA | AB672631.2 | - | 6 - 7 | 548 | 75 |
AB672632.1 | - | 6 - 7 | 550 | 75 | |
AB678452.1 | - | 5 - 6 | 625 | 75 | |
AB678453.1 | - | 5 - 6 | 477 | 75 | |
BC040679.2 | - | 5 - 6 | 548 | 75 | |
BX248046.1 | - | 4 - 5 | 421 | 75 | |
BX640825.1 | - | 5 - 6 | 872 | 75 | |
CN352234.1 | - | 5 - 6 | 495 | 75 |