Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010977.2 | NP_001010977.1 | 2 - 3 | 302 | 95 |
XM_017020407.1 | XP_016875896.1 | 3 - 4 | 1196 | 95 | |
XM_017020408.1 | XP_016875897.1 | 3 - 4 | 1749 | 95 | |
GenBank mRNA | BC132748.1 | - | 2 - 3 | 302 | 95 |
BC132750.1 | - | 2 - 3 | 302 | 95 |