Assay Design



Gene Transcripts

Gene Symbol
C11orf84
Entrez Gene ID
144097
Gene Name
chromosome 11 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.11: 63813374 - 63827718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138471.2 NP_612480.1 3 - 4 915 91
XM_005273782.2 XP_005273839.1 3 - 4 915 91
XM_005273783.2 XP_005273840.1 3 - 4 915 91
XM_006718437.1 XP_006718500.1 3 - 4 915 91
XM_011544770.1 XP_011543072.1 3 - 4 915 91
XM_011544772.1 XP_011543074.1 3 - 4 915 91
GenBank mRNA BC002782.2 - 3 - 4 673 91
BC007540.1 - 3 - 4 673 91
BC056402.1 - 3 - 4 838 91

View Full Product Details