Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027058.1 | 1 - 2 | 309 | 89 | |
GenBank mRNA | BX648413.1 | - | 1 - 2 | 269 | 89 |
DA068554.1 | - | 1 - 2 | 197 | 89 | |
DA916381.1 | - | 1 - 2 | 309 | 89 | |
FN357117.1 | - | 1 - 2 | 203 | 89 | |
FN357118.1 | - | 1 - 2 | 203 | 89 | |
FN357119.1 | - | 1 - 2 | 203 | 89 | |
FN357121.1 | - | 1 - 2 | 203 | 89 | |
FN357122.1 | - | 1 - 2 | 175 | 89 | |
FN357125.1 | - | 1 - 2 | 203 | 89 | |
FN357126.1 | - | 1 - 2 | 175 | 89 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204408.1 | NP_001191337.1 | 1 - 2 | 309 | 89 |
GenBank mRNA | FM995495.1 | - | 1 - 2 | 203 | 89 |
FM995497.1 | - | 1 - 2 | 203 | 89 |