Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015284.3 | NP_056099.3 | 11 - 12 | 1708 | 105 |
XM_005270686.3 | XP_005270743.1 | 12 - 13 | 1769 | 105 | |
XM_006710501.3 | XP_006710564.1 | 11 - 12 | 1656 | 105 | |
XM_011541106.2 | XP_011539408.1 | 11 - 12 | 1807 | 105 | |
XM_011541107.2 | XP_011539409.1 | 8 - 9 | 1304 | 105 | |
XM_017000819.1 | XP_016856308.1 | 12 - 13 | 1769 | 105 | |
XM_017000820.1 | XP_016856309.1 | 11 - 12 | 1598 | 105 | |
XM_017000821.1 | XP_016856310.1 | 8 - 9 | 1290 | 105 | |
GenBank mRNA | AK131107.1 | - | 2 - 3 | 355 | 105 |