Assay Design
Gene Transcripts
- Gene Symbol
- LINC00488
- Entrez Gene ID
-
677779
- Gene Name
- long intergenic non-protein coding RNA 488
- Gene Aliases
-
C3orf66
- Chromosome Location
-
Chr.3: 109178165 - 109185261 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026767.1 |
|
1 - 2 |
136 |
69 |
GenBank mRNA |
AK123226.1 |
- |
1 - 2 |
136 |
69 |
View Full Product Details