Assay Design



Gene Transcripts

Gene Symbol
WDR27
Entrez Gene ID
253769
Gene Name
WD repeat domain 27
Gene Aliases
-
Chromosome Location
Chr.6: 169426420 - 169702071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202550.1 NP_001189479.1 8 - 9 1339 80
NM_182552.4 NP_872358.4 11 - 12 1720 80
XM_011535682.2 XP_011533984.1 12 - 13 1910 80
XM_011535684.2 XP_011533986.1 12 - 13 1910 80
XM_011535685.2 XP_011533987.1 12 - 13 1910 80
XM_011535687.2 XP_011533989.1 12 - 13 1910 80
XM_011535688.2 XP_011533990.1 12 - 13 1910 80
XM_011535689.2 XP_011533991.1 12 - 13 1910 80
XM_011535690.2 XP_011533992.1 12 - 13 1910 80
XM_011535691.2 XP_011533993.1 12 - 13 1910 80
XM_011535692.2 XP_011533994.1 12 - 13 1910 80
XM_011535693.2 XP_011533995.1 12 - 13 1910 80
XM_011535694.2 XP_011533996.1 12 - 13 1910 80
XM_011535696.2 XP_011533998.1 12 - 13 1910 80
XM_011535697.2 XP_011533999.1 12 - 13 1910 80
XM_017010658.1 XP_016866147.1 12 - 13 1910 80
XM_017010659.1 XP_016866148.1 12 - 13 1910 80
XM_017010660.1 XP_016866149.1 12 - 13 1910 80
XM_017010661.1 XP_016866150.1 12 - 13 1910 80
XM_017010662.1 XP_016866151.1 12 - 13 1910 80
XM_017010663.1 XP_016866152.1 12 - 13 1910 80
XM_017010664.1 XP_016866153.1 12 - 13 1910 80
XM_017010665.1 XP_016866154.1 12 - 13 1910 80
XM_017010666.1 XP_016866155.1 12 - 13 1910 80
XM_017010667.1 XP_016866156.1 12 - 13 1910 80
XM_017010668.1 XP_016866157.1 12 - 13 1910 80
XM_017010669.1 XP_016866158.1 6 - 7 1155 80
XM_017010670.1 XP_016866159.1 12 - 13 1910 80
XM_017010671.1 XP_016866160.1 12 - 13 1910 80
XM_017010672.1 XP_016866161.1 12 - 13 1910 80
XM_017010673.1 XP_016866162.1 12 - 13 1910 80
XM_017010674.1 XP_016866163.1 11 - 12 1706 80
XM_017010675.1 XP_016866164.1 10 - 11 1524 80
GenBank mRNA AK128656.1 - - 2803 80
AK131435.1 - 2 - 3 2101 80
BC131633.1 - 8 - 9 862 80
BX648854.1 - 7 - 8 715 80

View Full Product Details