Assay Design
Gene Transcripts
- Gene Symbol
- COX5B
- Entrez Gene ID
-
1329
- Gene Name
- cytochrome c oxidase subunit 5B
- Gene Aliases
-
COXVB
- Chromosome Location
-
Chr.2: 97646058 - 97648194 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001862.2 |
NP_001853.2 |
1 - 2 |
129 |
146 |
GenBank mRNA |
BC006229.2 |
- |
1 - 2 |
129 |
146 |
View Full Product Details