Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003019.4 | NP_003010.4 | 2 - 3 | 237 | 102 |
XM_011540087.1 | XP_011538389.1 | 2 - 3 | 216 | 102 | |
XM_011540088.2 | XP_011538390.1 | 2 - 3 | 305 | 102 | |
GenBank mRNA | BC022318.1 | - | 2 - 3 | 238 | 102 |
BI820265.1 | - | 1 - 2 | 209 | 102 | |
CR541948.1 | - | 1 - 2 | 194 | 102 | |
X65018.1 | - | 2 - 3 | 365 | 102 |