Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006426.2 | NP_006417.2 | 3 - 4 | 469 | 68 |
XM_005252658.3 | XP_005252715.1 | 3 - 4 | 458 | 68 | |
XM_011539125.2 | XP_011537427.1 | 4 - 5 | 472 | 68 | |
XM_017015482.1 | XP_016870971.1 | 3 - 4 | 1733 | 68 | |
XM_017015483.1 | XP_016870972.1 | 4 - 5 | 1886 | 68 | |
GenBank mRNA | AB006713.1 | - | 3 - 4 | 463 | 68 |
BC029280.1 | - | 3 - 4 | 353 | 68 | |
BC136329.1 | - | 3 - 4 | 338 | 68 | |
BC136330.1 | - | 3 - 4 | 338 | 68 | |
Y10976.1 | - | 3 - 4 | 346 | 68 |