Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040458.1 | NP_001035548.1 | 14 - 15 | 2443 | 123 |
NM_001198541.1 | NP_001185470.1 | 14 - 15 | 2201 | 123 | |
NM_016442.3 | NP_057526.3 | 14 - 15 | 2443 | 123 | |
XM_005272015.4 | XP_005272072.1 | 14 - 15 | 2394 | 123 | |
XM_005272016.4 | XP_005272073.1 | 14 - 15 | 2354 | 123 | |
XM_011543480.2 | XP_011541782.1 | 18 - 19 | 4663 | 123 | |
XM_011543481.2 | XP_011541783.1 | 18 - 19 | 4660 | 123 | |
XM_011543484.2 | XP_011541786.1 | 18 - 19 | 4659 | 123 | |
XM_011543485.2 | XP_011541787.1 | 17 - 18 | 4479 | 123 | |
XM_011543486.2 | XP_011541788.1 | 18 - 19 | 4663 | 123 | |
XM_017009580.1 | XP_016865069.1 | 19 - 20 | 4719 | 123 | |
XM_017009581.1 | XP_016865070.1 | 17 - 18 | 4505 | 123 | |
XM_017009582.1 | XP_016865071.1 | 14 - 15 | 2178 | 123 | |
XM_017009583.1 | XP_016865072.1 | 14 - 15 | 2336 | 123 | |
GenBank mRNA | AB011097.2 | - | 13 - 14 | 2117 | 123 |
AF106037.1 | - | 14 - 15 | 2158 | 123 | |
AF183569.1 | - | 14 - 15 | 2163 | 123 | |
AF222340.1 | - | 14 - 15 | 2183 | 123 | |
AK291636.1 | - | 14 - 15 | 2193 | 123 | |
AK310535.1 | - | 13 - 14 | 1618 | 123 | |
AY028806.1 | - | 14 - 15 | 2193 | 123 | |
AY028807.1 | - | 14 - 15 | 2193 | 123 | |
AY358590.1 | - | 14 - 15 | 2181 | 123 | |
BC030775.1 | - | 14 - 15 | 2195 | 123 | |
KM357870.1 | - | 13 - 14 | 2096 | 123 | |
KM357871.1 | - | 13 - 14 | 2096 | 123 | |
KM357872.1 | - | 13 - 14 | 2096 | 123 | |
KM357873.1 | - | 13 - 14 | 2096 | 123 | |
KM357874.1 | - | 13 - 14 | 2096 | 123 | |
KM357875.1 | - | 13 - 14 | 2096 | 123 | |
KM357876.1 | - | 13 - 14 | 2096 | 123 | |
KM357877.1 | - | 13 - 14 | 2096 | 123 | |
KM357878.1 | - | 13 - 14 | 2096 | 123 | |
KM357879.1 | - | 13 - 14 | 2096 | 123 | |
KM357880.1 | - | 13 - 14 | 2096 | 123 | |
KM357881.1 | - | 13 - 14 | 2096 | 123 | |
KM357882.1 | - | 13 - 14 | 2096 | 123 | |
KM357883.1 | - | 13 - 14 | 2096 | 123 | |
KM357884.1 | - | 13 - 14 | 2096 | 123 | |
KM357885.1 | - | 13 - 14 | 2096 | 123 | |
KM357886.1 | - | 13 - 14 | 2096 | 123 | |
KM357887.1 | - | 13 - 14 | 2096 | 123 | |
KM357888.1 | - | 13 - 14 | 2096 | 123 | |
KM357889.1 | - | 13 - 14 | 2096 | 123 | |
KM357890.1 | - | 13 - 14 | 2096 | 123 | |
KM357891.1 | - | 13 - 14 | 2096 | 123 |