Assay Design



Gene Transcripts

Gene Symbol
CYP3A4
Entrez Gene ID
1576
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1
Chromosome Location
Chr.7: 99756960 - 99784188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202855.2 NP_001189784.1 8 - 9 907 92
NM_017460.5 NP_059488.2 8 - 9 910 92
XM_017011780.1 XP_016867269.1 8 - 9 908 92
GenBank mRNA AF182273.1 - 8 - 9 812 92
AK298451.1 - 7 - 8 781 92
AK298462.1 - 6 - 7 657 92
AK312967.1 - 8 - 9 903 92
AY606313.2 - 8 - 9 805 92
BC069418.1 - 8 - 9 853 92
BC101631.1 - 8 - 9 899 92
DQ924960.1 - 8 - 9 803 92
J04449.1 - 8 - 9 868 92
M14096.1 - 8 - 9 871 92
M18907.1 - 8 - 9 893 92
X12387.1 - 8 - 9 872 92

View Full Product Details