Assay Design
Gene Transcripts
- Gene Symbol
- MMP28
- Entrez Gene ID
-
79148
- Gene Name
- matrix metallopeptidase 28
- Gene Aliases
-
EPILYSIN, MM28, MMP-25, MMP-28, MMP25
- Chromosome Location
-
Chr.17: 35756249 - 35795707 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_032950.3 |
NP_116568.1 |
7 - 8 |
1494 |
85 |
GenBank mRNA |
BC002631.2 |
- |
7 - 8 |
1411 |
85 |
View Full Product Details