Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81702152-81702749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3412339 c.2105G>A p.R702H Chr.7: 81702663-81702663 on Build GRCh38
7371066 c.2186A>T p.*729L Chr.7: 81702582-81702582 on Build GRCh38
6969811 c.2074G>T p.V692F Chr.7: 81702694-81702694 on Build GRCh38
5873139 c.2029C>T p.L677F Chr.7: 81702739-81702739 on Build GRCh38
3030118 c.2104C>T p.R702C Chr.7: 81702664-81702664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992946888 Chr.7: 81702392-81702392 on Build GRCh38
rs899567940 Chr.7: 81702218-81702218 on Build GRCh38
rs563705580 Chr.7: 81702686-81702686 on Build GRCh38
rs140790665 Chr.7: 81702589-81702589 on Build GRCh38
rs182057547 Chr.7: 81702185-81702185 on Build GRCh38
rs764730119 Chr.7: 81702629-81702629 on Build GRCh38
rs374789745 Chr.7: 81702478-81702479 on Build GRCh38
rs573406409 Chr.7: 81702515-81702515 on Build GRCh38
rs890647502 Chr.7: 81702175-81702175 on Build GRCh38
rs1053369698 Chr.7: 81702331-81702331 on Build GRCh38
rs886062456 Chr.7: 81702480-81702480 on Build GRCh38
rs750104031 Chr.7: 81702698-81702698 on Build GRCh38
rs780528260 Chr.7: 81702583-81702583 on Build GRCh38
rs750490813 Chr.7: 81702533-81702533 on Build GRCh38
rs543827901 Chr.7: 81702684-81702684 on Build GRCh38
rs757972650 Chr.7: 81702581-81702581 on Build GRCh38
rs760728215 Chr.7: 81702658-81702658 on Build GRCh38
rs531222230 Chr.7: 81702413-81702413 on Build GRCh38
rs267601578 Chr.7: 81702638-81702638 on Build GRCh38
rs879724696 Chr.7: 81702238-81702238 on Build GRCh38
rs1051166277 Chr.7: 81702243-81702243 on Build GRCh38
rs977727635 Chr.7: 81702493-81702493 on Build GRCh38
rs539702697 Chr.7: 81702357-81702357 on Build GRCh38
rs994551612 Chr.7: 81702204-81702204 on Build GRCh38
rs1038792244 Chr.7: 81702187-81702187 on Build GRCh38
rs537468151 Chr.7: 81702228-81702228 on Build GRCh38
rs756957205 Chr.7: 81702559-81702559 on Build GRCh38
rs763933267 Chr.7: 81702663-81702663 on Build GRCh38
rs777071855 Chr.7: 81702599-81702599 on Build GRCh38
rs934877141 Chr.7: 81702330-81702330 on Build GRCh38
rs545984462 Chr.7: 81702264-81702267 on Build GRCh38
rs747516423 Chr.7: 81702585-81702585 on Build GRCh38
rs777313117 Chr.7: 81702322-81702322 on Build GRCh38
rs778483338 Chr.7: 81702571-81702571 on Build GRCh38
rs925862874 Chr.7: 81702569-81702569 on Build GRCh38
rs1034775459 Chr.7: 81702272-81702273 on Build GRCh38
rs878879830 Chr.7: 81702276-81702276 on Build GRCh38
rs960578259 Chr.7: 81702273-81702273 on Build GRCh38
rs763072412 Chr.7: 81702645-81702645 on Build GRCh38
rs568550366 Chr.7: 81702194-81702194 on Build GRCh38
rs1048682653 Chr.7: 81702562-81702562 on Build GRCh38
rs758013381 Chr.7: 81702309-81702309 on Build GRCh38
rs1004774004 Chr.7: 81702261-81702261 on Build GRCh38
rs775503666 Chr.7: 81702654-81702654 on Build GRCh38
rs1004721704 Chr.7: 81702576-81702576 on Build GRCh38
rs542402530 Chr.7: 81702524-81702524 on Build GRCh38
rs577625800 Chr.7: 81702315-81702315 on Build GRCh38
rs958744890 Chr.7: 81702379-81702379 on Build GRCh38
rs930299625 Chr.7: 81702399-81702399 on Build GRCh38
rs779001317 Chr.7: 81702570-81702573 on Build GRCh38
rs757849670 Chr.7: 81702709-81702709 on Build GRCh38
rs561484014 Chr.7: 81702602-81702602 on Build GRCh38
rs1027510708 Chr.7: 81702213-81702213 on Build GRCh38
rs993314088 Chr.7: 81702274-81702274 on Build GRCh38
rs201768315 Chr.7: 81702646-81702646 on Build GRCh38
rs763357973 Chr.7: 81702314-81702315 on Build GRCh38
rs886820277 Chr.7: 81702263-81702263 on Build GRCh38
rs900161616 Chr.7: 81702198-81702198 on Build GRCh38
rs186434043 Chr.7: 81702279-81702279 on Build GRCh38
rs753392862 Chr.7: 81702552-81702552 on Build GRCh38
rs765177187 Chr.7: 81702532-81702532 on Build GRCh38
rs377185441 Chr.7: 81702541-81702541 on Build GRCh38
rs767556298 Chr.7: 81702536-81702536 on Build GRCh38
rs933743740 Chr.7: 81702219-81702219 on Build GRCh38
rs745356078 Chr.7: 81702575-81702575 on Build GRCh38
rs767332482 Chr.7: 81702545-81702545 on Build GRCh38
rs544879896 Chr.7: 81702360-81702360 on Build GRCh38
rs770025635 Chr.7: 81702632-81702632 on Build GRCh38
rs1024724608 Chr.7: 81702422-81702422 on Build GRCh38
rs779568330 Chr.7: 81702727-81702727 on Build GRCh38
rs970443569 Chr.7: 81702313-81702313 on Build GRCh38
rs912015642 Chr.7: 81702351-81702351 on Build GRCh38
rs777717109 Chr.7: 81702550-81702550 on Build GRCh38
rs967428555 Chr.7: 81702462-81702462 on Build GRCh38
rs372366408 Chr.7: 81702673-81702673 on Build GRCh38

View Full Product Details