Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9739492-9739841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs149886683 Chr.X: 9739760-9739760 on Build GRCh38
rs756686806 Chr.X: 9739721-9739722 on Build GRCh38
rs776710609 Chr.X: 9739671-9739671 on Build GRCh38
rs768774548 Chr.X: 9739665-9739665 on Build GRCh38
rs756728406 Chr.X: 9739582-9739582 on Build GRCh38
rs140873054 Chr.X: 9739526-9739526 on Build GRCh38
rs766347844 Chr.X: 9739680-9739680 on Build GRCh38
rs748861491 Chr.X: 9739581-9739581 on Build GRCh38
rs771056170 Chr.X: 9739574-9739574 on Build GRCh38
rs745458128 Chr.X: 9739633-9739633 on Build GRCh38
rs776550135 Chr.X: 9739600-9739600 on Build GRCh38
rs968984180 Chr.X: 9739773-9739773 on Build GRCh38
rs759315924 Chr.X: 9739655-9739655 on Build GRCh38
rs763168997 Chr.X: 9739672-9739672 on Build GRCh38
rs138809166 Chr.X: 9739560-9739560 on Build GRCh38
rs1023047490 Chr.X: 9739572-9739572 on Build GRCh38
rs771928397 Chr.X: 9739654-9739654 on Build GRCh38
rs777471473 Chr.X: 9739550-9739550 on Build GRCh38
rs753117759 Chr.X: 9739756-9739756 on Build GRCh38
rs751704023 Chr.X: 9739554-9739554 on Build GRCh38
rs763113825 Chr.X: 9739546-9739546 on Build GRCh38
rs756548263 Chr.X: 9739571-9739571 on Build GRCh38
rs767931116 Chr.X: 9739753-9739753 on Build GRCh38
rs150179962 Chr.X: 9739527-9739527 on Build GRCh38
rs759639368 Chr.X: 9739699-9739699 on Build GRCh38
rs374388815 Chr.X: 9739530-9739530 on Build GRCh38
rs34324958 Chr.X: 9739569-9739569 on Build GRCh38
rs62635047 Chr.X: 9739673-9739674 on Build GRCh38
rs774656143 Chr.X: 9739692-9739692 on Build GRCh38
rs1030711663 Chr.X: 9739662-9739662 on Build GRCh38
rs769479021 Chr.X: 9739749-9739749 on Build GRCh38
rs954542195 Chr.X: 9739593-9739593 on Build GRCh38
rs866294644 Chr.X: 9739698-9739698 on Build GRCh38
rs773371296 Chr.X: 9739545-9739545 on Build GRCh38
rs746959492 Chr.X: 9739660-9739660 on Build GRCh38
rs755421333 Chr.X: 9739555-9739555 on Build GRCh38
rs980350774 Chr.X: 9739796-9739796 on Build GRCh38

View Full Product Details