Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 49 - 50 | 7048 | 91 |
XM_005251773.2 | XP_005251830.1 | 49 - 50 | 6963 | 91 | |
XM_005251776.2 | XP_005251833.1 | 48 - 49 | 6803 | 91 | |
XM_011518339.2 | XP_011516641.1 | 50 - 51 | 7038 | 91 | |
XM_011518340.2 | XP_011516642.1 | 51 - 52 | 7253 | 91 | |
XM_011518341.2 | XP_011516643.1 | 50 - 51 | 7032 | 91 | |
XM_011518342.2 | XP_011516644.1 | 47 - 48 | 6741 | 91 | |
XM_017014378.1 | XP_016869867.1 | 50 - 51 | 7236 | 91 | |
XM_017014379.1 | XP_016869868.1 | 50 - 51 | 7644 | 91 | |
XM_017014380.1 | XP_016869869.1 | 50 - 51 | 6970 | 91 | |
XM_017014381.1 | XP_016869870.1 | 50 - 51 | 7036 | 91 | |
XM_017014382.1 | XP_016869871.1 | 48 - 49 | 7501 | 91 | |
GenBank mRNA | AB055982.1 | - | 48 - 49 | 6644 | 91 |
AB208839.1 | - | 10 - 11 | 3684 | 91 | |
AB445477.1 | - | 48 - 49 | 6644 | 91 | |
AF165281.1 | - | 47 - 48 | 6584 | 91 | |
AF285167.1 | - | 49 - 50 | 6934 | 91 | |
AJ012376.1 | - | 47 - 48 | 6584 | 91 | |
AK027864.1 | - | 14 - 15 | 1771 | 91 | |
BC146856.1 | - | 49 - 50 | 6858 | 91 |