Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226891558-226892063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
679334 c.1063G>A p.E355K Chr.1: 226891835-226891835 on Build GRCh38
4519497 c.1111G>A p.E371K Chr.1: 226891784-226891784 on Build GRCh38
1689923 c.1010_1011CC>AT p.P337H Chr.1: 226891782-226891783 on Build GRCh38
6757463 c.1158delG p.E387fs*7 Chr.1: 226891831-226891831 on Build GRCh38
6757462 c.1059delG p.E354fs*7 Chr.1: 226891831-226891831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765039787 Chr.1: 226891755-226891756 on Build GRCh38
rs753933727 Chr.1: 226891732-226891732 on Build GRCh38
rs534507840 Chr.1: 226891980-226891980 on Build GRCh38
rs767982599 Chr.1: 226891716-226891716 on Build GRCh38
rs12130732 Chr.1: 226891999-226891999 on Build GRCh38
rs898967639 Chr.1: 226891739-226891739 on Build GRCh38
rs991591942 Chr.1: 226891766-226891766 on Build GRCh38
rs987514889 Chr.1: 226891578-226891578 on Build GRCh38
rs779618448 Chr.1: 226891879-226891879 on Build GRCh38
rs924766442 Chr.1: 226891857-226891857 on Build GRCh38
rs375520284 Chr.1: 226891885-226891885 on Build GRCh38
rs774972844 Chr.1: 226891714-226891714 on Build GRCh38
rs765420103 Chr.1: 226891824-226891824 on Build GRCh38
rs900945278 Chr.1: 226891918-226891918 on Build GRCh38
rs968604078 Chr.1: 226891865-226891865 on Build GRCh38
rs63750207 Chr.1: 226891773-226891773 on Build GRCh38
rs760890655 Chr.1: 226891725-226891725 on Build GRCh38
rs1056851112 Chr.1: 226891689-226891691 on Build GRCh38
rs942398013 Chr.1: 226891886-226891886 on Build GRCh38
rs375609936 Chr.1: 226891699-226891699 on Build GRCh38
rs572118237 Chr.1: 226891940-226891940 on Build GRCh38
rs750726916 Chr.1: 226891719-226891719 on Build GRCh38
rs1003275521 Chr.1: 226891904-226891904 on Build GRCh38
rs751424049 Chr.1: 226891861-226891861 on Build GRCh38
rs755095033 Chr.1: 226891741-226891741 on Build GRCh38
rs200044441 Chr.1: 226891765-226891765 on Build GRCh38
rs760977533 Chr.1: 226891801-226891801 on Build GRCh38
rs370856356 Chr.1: 226892001-226892001 on Build GRCh38
rs202142711 Chr.1: 226891891-226891891 on Build GRCh38
rs997925982 Chr.1: 226891919-226891919 on Build GRCh38
rs541841648 Chr.1: 226891586-226891586 on Build GRCh38
rs201917728 Chr.1: 226891787-226891787 on Build GRCh38
rs548161965 Chr.1: 226891866-226891866 on Build GRCh38
rs199881176 Chr.1: 226891795-226891795 on Build GRCh38
rs372224790 Chr.1: 226891892-226891892 on Build GRCh38
rs758364204 Chr.1: 226891849-226891849 on Build GRCh38
rs951299192 Chr.1: 226891665-226891665 on Build GRCh38
rs114127255 Chr.1: 226891972-226891972 on Build GRCh38
rs769386876 Chr.1: 226891783-226891783 on Build GRCh38
rs75612539 Chr.1: 226892013-226892013 on Build GRCh38
rs1040287058 Chr.1: 226892002-226892002 on Build GRCh38
rs565448044 Chr.1: 226891671-226891671 on Build GRCh38
rs78420366 Chr.1: 226891821-226891821 on Build GRCh38
rs1035731961 Chr.1: 226891926-226891926 on Build GRCh38
rs758259518 Chr.1: 226891769-226891769 on Build GRCh38
rs988194633 Chr.1: 226891605-226891605 on Build GRCh38
rs769066300 Chr.1: 226891713-226891713 on Build GRCh38
rs766544235 Chr.1: 226891730-226891730 on Build GRCh38
rs368035858 Chr.1: 226891874-226891874 on Build GRCh38
rs377406802 Chr.1: 226891692-226891692 on Build GRCh38
rs759669954 Chr.1: 226891817-226891817 on Build GRCh38
rs971912924 Chr.1: 226892022-226892022 on Build GRCh38
rs773231811 Chr.1: 226892041-226892041 on Build GRCh38
rs745572658 Chr.1: 226891869-226891869 on Build GRCh38
rs773597667 Chr.1: 226891796-226891796 on Build GRCh38
rs199854075 Chr.1: 226891706-226891706 on Build GRCh38
rs764086543 Chr.1: 226891855-226891855 on Build GRCh38
rs766705508 Chr.1: 226891813-226891813 on Build GRCh38
rs199937781 Chr.1: 226891902-226891902 on Build GRCh38
rs777639021 Chr.1: 226891772-226891772 on Build GRCh38
rs537527025 Chr.1: 226891900-226891900 on Build GRCh38
rs776874000 Chr.1: 226891814-226891814 on Build GRCh38
rs112130067 Chr.1: 226891691-226891693 on Build GRCh38
rs1036103304 Chr.1: 226891957-226891957 on Build GRCh38
rs956708647 Chr.1: 226891808-226891808 on Build GRCh38
rs915371102 Chr.1: 226891633-226891633 on Build GRCh38
rs559617407 Chr.1: 226891784-226891784 on Build GRCh38
rs116635380 Chr.1: 226891942-226891942 on Build GRCh38
rs372305083 Chr.1: 226891882-226891882 on Build GRCh38
rs762769497 Chr.1: 226891898-226891899 on Build GRCh38
rs771938561 Chr.1: 226891697-226891697 on Build GRCh38
rs200691879 Chr.1: 226891703-226891703 on Build GRCh38
rs543669793 Chr.1: 226891619-226891619 on Build GRCh38
rs757236307 Chr.1: 226891672-226891672 on Build GRCh38
rs946924222 Chr.1: 226891639-226891639 on Build GRCh38
rs773094480 Chr.1: 226891841-226891842 on Build GRCh38
rs1050313305 Chr.1: 226891991-226891991 on Build GRCh38
rs778824612 Chr.1: 226891745-226891745 on Build GRCh38
rs200977587 Chr.1: 226891840-226891840 on Build GRCh38
rs575395926 Chr.1: 226891601-226891601 on Build GRCh38
rs535203364 Chr.1: 226891682-226891682 on Build GRCh38
rs781365270 Chr.1: 226891777-226891777 on Build GRCh38
rs574523822 Chr.1: 226891577-226891577 on Build GRCh38
rs1018984857 Chr.1: 226891591-226891591 on Build GRCh38
rs776289096 Chr.1: 226891707-226891707 on Build GRCh38
rs535474711 Chr.1: 226892035-226892035 on Build GRCh38
rs530065304 Chr.1: 226891786-226891786 on Build GRCh38
rs984012936 Chr.1: 226891687-226891687 on Build GRCh38
rs202106330 Chr.1: 226891883-226891883 on Build GRCh38
rs759079072 Chr.1: 226891709-226891709 on Build GRCh38
rs886910587 Chr.1: 226891963-226891963 on Build GRCh38
rs994711686 Chr.1: 226891754-226891754 on Build GRCh38
rs371097422 Chr.1: 226891694-226891694 on Build GRCh38
rs757169630 Chr.1: 226891862-226891862 on Build GRCh38
rs373934941 Chr.1: 226891764-226891764 on Build GRCh38
rs7537037 Chr.1: 226891600-226891600 on Build GRCh38
rs532697278 Chr.1: 226891690-226891690 on Build GRCh38
rs1013367655 Chr.1: 226892005-226892005 on Build GRCh38
rs747956848 Chr.1: 226892000-226892000 on Build GRCh38
rs201663855 Chr.1: 226891715-226891715 on Build GRCh38

View Full Product Details