Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25880954-25881539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
UniGene
Hs.434980
Gene Chromosome Location
Chr.21: 25880550-26171128 on Build GRCh38
Transcripts
NM_000484.3 NM_001136016.3 NM_001136129.2 NM_001136130.2 NM_001136131.2 NM_001204301.1 NM_001204302.1 NM_001204303.1 NM_201413.2 NM_201414.2

SNP Detail

SNP ID Chromosome Location
rs930856936 Chr.21: 25881152-25881153 on Build GRCh38
rs1031961200 Chr.21: 25881483-25881483 on Build GRCh38
rs748281356 Chr.21: 25881079-25881079 on Build GRCh38
rs201853089 Chr.21: 25881380-25881380 on Build GRCh38
rs780350314 Chr.21: 25881493-25881493 on Build GRCh38
rs201034704 Chr.21: 25881087-25881087 on Build GRCh38
rs950281598 Chr.21: 25881469-25881470 on Build GRCh38
rs45455403 Chr.21: 25881080-25881080 on Build GRCh38
rs898132746 Chr.21: 25881013-25881013 on Build GRCh38
rs150007519 Chr.21: 25881259-25881259 on Build GRCh38
rs532399598 Chr.21: 25881263-25881263 on Build GRCh38
rs966712982 Chr.21: 25881013-25881013 on Build GRCh38
rs906248824 Chr.21: 25881199-25881199 on Build GRCh38
rs983695777 Chr.21: 25881444-25881444 on Build GRCh38
rs201729905 Chr.21: 25881406-25881406 on Build GRCh38
rs200600383 Chr.21: 25881283-25881283 on Build GRCh38
rs201380777 Chr.21: 25881470-25881470 on Build GRCh38
rs970148921 Chr.21: 25881506-25881506 on Build GRCh38
rs1010797044 Chr.21: 25880987-25880987 on Build GRCh38
rs901984087 Chr.21: 25881452-25881452 on Build GRCh38
rs568756389 Chr.21: 25881399-25881399 on Build GRCh38
rs201624429 Chr.21: 25881236-25881236 on Build GRCh38
rs889532960 Chr.21: 25881064-25881064 on Build GRCh38
rs1011605214 Chr.21: 25881495-25881495 on Build GRCh38
rs139278773 Chr.21: 25881067-25881067 on Build GRCh38
rs200591716 Chr.21: 25881396-25881396 on Build GRCh38
rs201377891 Chr.21: 25881291-25881291 on Build GRCh38
rs966680337 Chr.21: 25881253-25881253 on Build GRCh38
rs1019370324 Chr.21: 25881349-25881349 on Build GRCh38
rs11549660 Chr.21: 25881020-25881020 on Build GRCh38
rs1048029462 Chr.21: 25881153-25881153 on Build GRCh38
rs572356515 Chr.21: 25881007-25881007 on Build GRCh38
rs199977643 Chr.21: 25881476-25881476 on Build GRCh38
rs919590033 Chr.21: 25881360-25881360 on Build GRCh38
rs200981079 Chr.21: 25881208-25881208 on Build GRCh38
rs1020893568 Chr.21: 25881012-25881012 on Build GRCh38
rs201644219 Chr.21: 25881472-25881472 on Build GRCh38
rs199862130 Chr.21: 25881181-25881181 on Build GRCh38
rs200058308 Chr.21: 25881407-25881407 on Build GRCh38
rs551830226 Chr.21: 25881335-25881335 on Build GRCh38
rs908163637 Chr.21: 25881454-25881454 on Build GRCh38
rs908194392 Chr.21: 25881161-25881164 on Build GRCh38
rs957353499 Chr.21: 25881237-25881237 on Build GRCh38
rs199680232 Chr.21: 25881046-25881046 on Build GRCh38
rs573263845 Chr.21: 25881234-25881234 on Build GRCh38
rs981297249 Chr.21: 25881297-25881297 on Build GRCh38
rs202194773 Chr.21: 25881210-25881210 on Build GRCh38
rs201876780 Chr.21: 25881499-25881499 on Build GRCh38
rs987511254 Chr.21: 25881016-25881020 on Build GRCh38
rs755797592 Chr.21: 25881357-25881357 on Build GRCh38
rs201887246 Chr.21: 25881156-25881156 on Build GRCh38
rs199765959 Chr.21: 25881266-25881266 on Build GRCh38
rs200736438 Chr.21: 25881299-25881299 on Build GRCh38
rs964795018 Chr.21: 25881350-25881350 on Build GRCh38
rs952218766 Chr.21: 25881442-25881442 on Build GRCh38
rs182834675 Chr.21: 25881196-25881196 on Build GRCh38
rs201008117 Chr.21: 25881052-25881052 on Build GRCh38
rs1054943545 Chr.21: 25881201-25881201 on Build GRCh38
rs931066255 Chr.21: 25881032-25881032 on Build GRCh38
rs999033118 Chr.21: 25881192-25881192 on Build GRCh38
rs912021116 Chr.21: 25881029-25881029 on Build GRCh38
rs943764960 Chr.21: 25881434-25881434 on Build GRCh38
rs754292576 Chr.21: 25881092-25881093 on Build GRCh38
rs200492624 Chr.21: 25881216-25881216 on Build GRCh38
rs558948870 Chr.21: 25881260-25881260 on Build GRCh38
rs975249008 Chr.21: 25881365-25881368 on Build GRCh38
rs532876832 Chr.21: 25881337-25881337 on Build GRCh38
rs201961275 Chr.21: 25880993-25880993 on Build GRCh38
rs970533877 Chr.21: 25881286-25881286 on Build GRCh38
rs1032097030 Chr.21: 25881209-25881209 on Build GRCh38
rs187940037 Chr.21: 25881298-25881298 on Build GRCh38
rs1040531485 Chr.21: 25881436-25881436 on Build GRCh38
rs199534609 Chr.21: 25881381-25881381 on Build GRCh38
rs200917677 Chr.21: 25881492-25881492 on Build GRCh38
rs939934369 Chr.21: 25881315-25881315 on Build GRCh38
rs1046036943 Chr.21: 25881471-25881471 on Build GRCh38
rs191651536 Chr.21: 25881112-25881112 on Build GRCh38
rs200262591 Chr.21: 25881256-25881256 on Build GRCh38
rs889564811 Chr.21: 25881385-25881385 on Build GRCh38
rs1046568428 Chr.21: 25881186-25881186 on Build GRCh38
rs200980048 Chr.21: 25881219-25881219 on Build GRCh38
rs1020864110 Chr.21: 25881251-25881251 on Build GRCh38

View Full Product Details