Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6021556-6022002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239126 c.499G>A p.V167I Chr.10: 6021562-6021562 on Build GRCh38
5599697 c.464G>A p.G155E Chr.10: 6021597-6021597 on Build GRCh38
143395 c.392_393GG>AA p.W131* Chr.10: 6021668-6021669 on Build GRCh38
5796878 c.409G>C p.E137Q Chr.10: 6021652-6021652 on Build GRCh38
4491758 c.385C>T p.P129S Chr.10: 6021676-6021676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012772481 Chr.10: 6021790-6021790 on Build GRCh38
rs766504939 Chr.10: 6021636-6021636 on Build GRCh38
rs201188114 Chr.10: 6021604-6021604 on Build GRCh38
rs760320762 Chr.10: 6021981-6021981 on Build GRCh38
rs773829518 Chr.10: 6021583-6021583 on Build GRCh38
rs1020018735 Chr.10: 6021945-6021945 on Build GRCh38
rs961416919 Chr.10: 6021848-6021848 on Build GRCh38
rs995990945 Chr.10: 6021910-6021910 on Build GRCh38
rs887140109 Chr.10: 6021597-6021597 on Build GRCh38
rs942200 Chr.10: 6021711-6021711 on Build GRCh38
rs763862564 Chr.10: 6021726-6021726 on Build GRCh38
rs377104035 Chr.10: 6021605-6021605 on Build GRCh38
rs374735363 Chr.10: 6021732-6021732 on Build GRCh38
rs779802160 Chr.10: 6021670-6021670 on Build GRCh38
rs141246728 Chr.10: 6021576-6021576 on Build GRCh38
rs902887175 Chr.10: 6021932-6021932 on Build GRCh38
rs763915787 Chr.10: 6021635-6021635 on Build GRCh38
rs976874957 Chr.10: 6021852-6021852 on Build GRCh38
rs763312995 Chr.10: 6021633-6021633 on Build GRCh38
rs772585400 Chr.10: 6021582-6021582 on Build GRCh38
rs767446188 Chr.10: 6021737-6021737 on Build GRCh38
rs747351412 Chr.10: 6021679-6021679 on Build GRCh38
rs1004799640 Chr.10: 6021600-6021600 on Build GRCh38
rs750345445 Chr.10: 6021738-6021738 on Build GRCh38
rs996425446 Chr.10: 6021607-6021607 on Build GRCh38
rs141655051 Chr.10: 6021625-6021625 on Build GRCh38
rs191340172 Chr.10: 6021709-6021709 on Build GRCh38
rs899080711 Chr.10: 6021908-6021908 on Build GRCh38
rs966986357 Chr.10: 6021960-6021960 on Build GRCh38
rs375013777 Chr.10: 6021873-6021873 on Build GRCh38
rs36086043 Chr.10: 6021867-6021867 on Build GRCh38
rs1056837693 Chr.10: 6021979-6021979 on Build GRCh38
rs546465474 Chr.10: 6021650-6021650 on Build GRCh38
rs745306282 Chr.10: 6021657-6021657 on Build GRCh38
rs12722695 Chr.10: 6021800-6021800 on Build GRCh38
rs755713136 Chr.10: 6021668-6021668 on Build GRCh38
rs765352295 Chr.10: 6021712-6021712 on Build GRCh38
rs959898602 Chr.10: 6021785-6021785 on Build GRCh38
rs763226676 Chr.10: 6021720-6021720 on Build GRCh38
rs773687056 Chr.10: 6021677-6021677 on Build GRCh38
rs761314426 Chr.10: 6021593-6021593 on Build GRCh38
rs182768172 Chr.10: 6021739-6021739 on Build GRCh38
rs781005101 Chr.10: 6021651-6021651 on Build GRCh38
rs772563186 Chr.10: 6021676-6021676 on Build GRCh38
rs751445826 Chr.10: 6021644-6021644 on Build GRCh38
rs1035484482 Chr.10: 6021748-6021748 on Build GRCh38
rs41310223 Chr.10: 6021879-6021879 on Build GRCh38
rs371733805 Chr.10: 6021740-6021740 on Build GRCh38
rs1015601894 Chr.10: 6021840-6021840 on Build GRCh38
rs765038171 Chr.10: 6021742-6021742 on Build GRCh38
rs12722694 Chr.10: 6021821-6021821 on Build GRCh38
rs747363078 Chr.10: 6021745-6021745 on Build GRCh38
rs147975326 Chr.10: 6021622-6021622 on Build GRCh38
rs766358118 Chr.10: 6021602-6021602 on Build GRCh38
rs765403608 Chr.10: 6021608-6021608 on Build GRCh38
rs778438730 Chr.10: 6021743-6021743 on Build GRCh38
rs886047082 Chr.10: 6021634-6021634 on Build GRCh38
rs748938194 Chr.10: 6021741-6021741 on Build GRCh38
rs575199237 Chr.10: 6021620-6021620 on Build GRCh38
rs759063933 Chr.10: 6021839-6021839 on Build GRCh38
rs562865947 Chr.10: 6021872-6021872 on Build GRCh38
rs146966522 Chr.10: 6021577-6021577 on Build GRCh38
rs774758997 Chr.10: 6021811-6021811 on Build GRCh38
rs267602533 Chr.10: 6021640-6021640 on Build GRCh38
rs757007076 Chr.10: 6021734-6021734 on Build GRCh38
rs12722693 Chr.10: 6021881-6021881 on Build GRCh38
rs767036534 Chr.10: 6021923-6021923 on Build GRCh38
rs775282989 Chr.10: 6021581-6021581 on Build GRCh38
rs934402658 Chr.10: 6021956-6021956 on Build GRCh38
rs1050040943 Chr.10: 6021664-6021664 on Build GRCh38
rs79887257 Chr.10: 6021931-6021931 on Build GRCh38

View Full Product Details