Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85980961-85981228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5641750 c.1684G>T p.V562L Chr.11: 85981224-85981224 on Build GRCh38
5373750 c.1740A>T p.P580P Chr.11: 85981168-85981168 on Build GRCh38
7214475 c.1770T>G p.A590A Chr.11: 85981138-85981138 on Build GRCh38
5641751 c.1534G>T p.V512L Chr.11: 85981224-85981224 on Build GRCh38
3953880 c.1536A>C p.V512V Chr.11: 85981222-85981222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750877404 Chr.11: 85981115-85981116 on Build GRCh38
rs1002533122 Chr.11: 85981051-85981051 on Build GRCh38
rs780443484 Chr.11: 85981082-85981082 on Build GRCh38
rs774673853 Chr.11: 85981011-85981011 on Build GRCh38
rs935373631 Chr.11: 85981115-85981115 on Build GRCh38
rs694353 Chr.11: 85981138-85981138 on Build GRCh38
rs779071423 Chr.11: 85981178-85981178 on Build GRCh38
rs1043858 Chr.11: 85981174-85981174 on Build GRCh38
rs1043859 Chr.11: 85981173-85981173 on Build GRCh38
rs142681055 Chr.11: 85981067-85981067 on Build GRCh38
rs780780083 Chr.11: 85981146-85981147 on Build GRCh38
rs764924889 Chr.11: 85981144-85981144 on Build GRCh38
rs200817695 Chr.11: 85981157-85981157 on Build GRCh38
rs768858872 Chr.11: 85981090-85981090 on Build GRCh38
rs982242065 Chr.11: 85981106-85981106 on Build GRCh38
rs752421488 Chr.11: 85981149-85981149 on Build GRCh38
rs766288437 Chr.11: 85981137-85981137 on Build GRCh38
rs1047877212 Chr.11: 85980999-85980999 on Build GRCh38
rs773368811 Chr.11: 85981109-85981109 on Build GRCh38
rs763666917 Chr.11: 85981153-85981153 on Build GRCh38
rs371902776 Chr.11: 85981094-85981094 on Build GRCh38
rs970048787 Chr.11: 85981050-85981050 on Build GRCh38
rs377370233 Chr.11: 85981167-85981167 on Build GRCh38
rs755346304 Chr.11: 85981177-85981177 on Build GRCh38
rs17817690 Chr.11: 85981018-85981018 on Build GRCh38
rs12295435 Chr.11: 85981150-85981150 on Build GRCh38
rs377101068 Chr.11: 85981131-85981131 on Build GRCh38
rs749670107 Chr.11: 85981086-85981086 on Build GRCh38
rs374615922 Chr.11: 85981164-85981164 on Build GRCh38
rs758985249 Chr.11: 85981143-85981144 on Build GRCh38
rs773209888 Chr.11: 85981127-85981127 on Build GRCh38
rs151012359 Chr.11: 85981113-85981113 on Build GRCh38
rs987918398 Chr.11: 85981111-85981111 on Build GRCh38
rs748164441 Chr.11: 85981099-85981099 on Build GRCh38
rs139710547 Chr.11: 85981034-85981034 on Build GRCh38
rs187292071 Chr.11: 85980991-85980991 on Build GRCh38
rs751078013 Chr.11: 85981155-85981155 on Build GRCh38

View Full Product Details