Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161349635-161350204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6547867 c.1363C>A p.R455S Chr.6: 161350134-161350134 on Build GRCh38
5019623 c.1310C>T p.P437L Chr.6: 161350187-161350187 on Build GRCh38
3024329 c.1311G>A p.P437P Chr.6: 161350186-161350186 on Build GRCh38
3782024 c.1391A>T p.D464V Chr.6: 161350106-161350106 on Build GRCh38
6932924 c.1341G>A p.W447* Chr.6: 161350156-161350156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965019796 Chr.6: 161349926-161349926 on Build GRCh38
rs537129880 Chr.6: 161349856-161349856 on Build GRCh38
rs942502004 Chr.6: 161350135-161350135 on Build GRCh38
rs893623989 Chr.6: 161349831-161349831 on Build GRCh38
rs764753874 Chr.6: 161349727-161349727 on Build GRCh38
rs772592654 Chr.6: 161350167-161350167 on Build GRCh38
rs886061236 Chr.6: 161349785-161349785 on Build GRCh38
rs781566147 Chr.6: 161349992-161349993 on Build GRCh38
rs940809817 Chr.6: 161349705-161349705 on Build GRCh38
rs779721381 Chr.6: 161350132-161350132 on Build GRCh38
rs368253350 Chr.6: 161350091-161350091 on Build GRCh38
rs62637702 Chr.6: 161350005-161350005 on Build GRCh38
rs544295360 Chr.6: 161350158-161350158 on Build GRCh38
rs779138830 Chr.6: 161350117-161350118 on Build GRCh38
rs182893847 Chr.6: 161350125-161350125 on Build GRCh38
rs755627153 Chr.6: 161350131-161350131 on Build GRCh38
rs777985497 Chr.6: 161349986-161349986 on Build GRCh38
rs908339150 Chr.6: 161350152-161350152 on Build GRCh38
rs748955949 Chr.6: 161350133-161350133 on Build GRCh38
rs35125035 Chr.6: 161350084-161350084 on Build GRCh38
rs751485437 Chr.6: 161350121-161350121 on Build GRCh38
rs998221207 Chr.6: 161349929-161349929 on Build GRCh38
rs768939316 Chr.6: 161350062-161350062 on Build GRCh38
rs763854303 Chr.6: 161350103-161350103 on Build GRCh38
rs916630868 Chr.6: 161350003-161350003 on Build GRCh38
rs200940812 Chr.6: 161350050-161350050 on Build GRCh38
rs755354082 Chr.6: 161350143-161350143 on Build GRCh38
rs543236357 Chr.6: 161349977-161349977 on Build GRCh38
rs771094906 Chr.6: 161350001-161350001 on Build GRCh38
rs772078706 Chr.6: 161350075-161350075 on Build GRCh38
rs748617874 Chr.6: 161350159-161350159 on Build GRCh38
rs573670651 Chr.6: 161349993-161349993 on Build GRCh38
rs747153578 Chr.6: 161349816-161349816 on Build GRCh38
rs1048255222 Chr.6: 161349661-161349661 on Build GRCh38
rs748710039 Chr.6: 161350071-161350071 on Build GRCh38
rs896441226 Chr.6: 161350000-161350000 on Build GRCh38
rs778305273 Chr.6: 161350176-161350176 on Build GRCh38
rs544293869 Chr.6: 161349991-161349992 on Build GRCh38
rs61730194 Chr.6: 161350083-161350083 on Build GRCh38
rs1016547536 Chr.6: 161349992-161349992 on Build GRCh38
rs892646687 Chr.6: 161350053-161350053 on Build GRCh38
rs763420602 Chr.6: 161349835-161349835 on Build GRCh38
rs1025324736 Chr.6: 161349936-161349936 on Build GRCh38
rs576586040 Chr.6: 161349798-161349798 on Build GRCh38
rs975260598 Chr.6: 161349938-161349938 on Build GRCh38
rs1019668090 Chr.6: 161349916-161349916 on Build GRCh38
rs774913629 Chr.6: 161350068-161350068 on Build GRCh38
rs545573732 Chr.6: 161350007-161350007 on Build GRCh38
rs747064211 Chr.6: 161350178-161350178 on Build GRCh38
rs965138593 Chr.6: 161350086-161350086 on Build GRCh38
rs137853056 Chr.6: 161350139-161350139 on Build GRCh38
rs1036457508 Chr.6: 161349997-161349997 on Build GRCh38
rs935354493 Chr.6: 161350046-161350046 on Build GRCh38
rs757263675 Chr.6: 161350123-161350123 on Build GRCh38
rs779799813 Chr.6: 161349749-161349749 on Build GRCh38
rs749641579 Chr.6: 161350060-161350060 on Build GRCh38
rs553007666 Chr.6: 161350039-161350039 on Build GRCh38
rs575581029 Chr.6: 161349983-161349984 on Build GRCh38
rs997369852 Chr.6: 161350101-161350101 on Build GRCh38
rs984344247 Chr.6: 161349991-161349991 on Build GRCh38
rs949363905 Chr.6: 161350004-161350004 on Build GRCh38
rs905483638 Chr.6: 161349785-161349785 on Build GRCh38
rs534613701 Chr.6: 161349761-161349761 on Build GRCh38
rs940743180 Chr.6: 161349996-161349996 on Build GRCh38
rs1013811412 Chr.6: 161349782-161349782 on Build GRCh38
rs961239925 Chr.6: 161350163-161350163 on Build GRCh38
rs949516613 Chr.6: 161350004-161350005 on Build GRCh38
rs956639141 Chr.6: 161349880-161349880 on Build GRCh38
rs756738207 Chr.6: 161349694-161349694 on Build GRCh38
rs71653628 Chr.6: 161349779-161349779 on Build GRCh38
rs767529432 Chr.6: 161350014-161350014 on Build GRCh38
rs776761587 Chr.6: 161350089-161350089 on Build GRCh38
rs1032542904 Chr.6: 161349800-161349800 on Build GRCh38
rs557142572 Chr.6: 161349978-161349978 on Build GRCh38
rs767967502 Chr.6: 161349993-161349994 on Build GRCh38
rs1023897256 Chr.6: 161349783-161349783 on Build GRCh38
rs758368939 Chr.6: 161349729-161349729 on Build GRCh38
rs1053659582 Chr.6: 161349812-161349812 on Build GRCh38
rs112022904 Chr.6: 161350097-161350097 on Build GRCh38
rs942203127 Chr.6: 161349665-161349665 on Build GRCh38
rs987924109 Chr.6: 161349909-161349909 on Build GRCh38
rs951001614 Chr.6: 161349945-161349945 on Build GRCh38

View Full Product Details