Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25880459-25881055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
UniGene
Hs.434980
Gene Chromosome Location
Chr.21: 25880550-26171128 on Build GRCh38
Transcripts
NM_000484.3 NM_001136016.3 NM_001136129.2 NM_001136130.2 NM_001136131.2 NM_001204301.1 NM_001204302.1 NM_001204303.1 NM_201413.2 NM_201414.2

SNP Detail

SNP ID Chromosome Location
rs572241749 Chr.21: 25880708-25880708 on Build GRCh38
rs543194262 Chr.21: 25880786-25880786 on Build GRCh38
rs528792746 Chr.21: 25880874-25880874 on Build GRCh38
rs995212130 Chr.21: 25880688-25880688 on Build GRCh38
rs201477224 Chr.21: 25880530-25880530 on Build GRCh38
rs202141287 Chr.21: 25880705-25880705 on Build GRCh38
rs200665110 Chr.21: 25880975-25880975 on Build GRCh38
rs1010797044 Chr.21: 25880987-25880987 on Build GRCh38
rs146265874 Chr.21: 25880525-25880525 on Build GRCh38
rs915716708 Chr.21: 25880685-25880685 on Build GRCh38
rs768025228 Chr.21: 25880948-25880948 on Build GRCh38
rs867380299 Chr.21: 25880915-25880915 on Build GRCh38
rs913917968 Chr.21: 25880794-25880794 on Build GRCh38
rs899374269 Chr.21: 25880845-25880845 on Build GRCh38
rs202132811 Chr.21: 25880935-25880935 on Build GRCh38
rs201838604 Chr.21: 25880567-25880567 on Build GRCh38
rs202081585 Chr.21: 25880757-25880757 on Build GRCh38
rs200853376 Chr.21: 25880852-25880852 on Build GRCh38
rs1025204856 Chr.21: 25880731-25880731 on Build GRCh38
rs190880668 Chr.21: 25880802-25880802 on Build GRCh38
rs549234553 Chr.21: 25880826-25880826 on Build GRCh38
rs1019197390 Chr.21: 25880814-25880814 on Build GRCh38
rs201019734 Chr.21: 25880646-25880646 on Build GRCh38
rs1039318980 Chr.21: 25880590-25880590 on Build GRCh38
rs1033939405 Chr.21: 25880947-25880947 on Build GRCh38
rs573994403 Chr.21: 25880673-25880673 on Build GRCh38
rs537547227 Chr.21: 25880913-25880913 on Build GRCh38
rs986373038 Chr.21: 25880568-25880568 on Build GRCh38
rs201611727 Chr.21: 25880807-25880807 on Build GRCh38
rs987511254 Chr.21: 25881016-25881020 on Build GRCh38
rs183019783 Chr.21: 25880894-25880894 on Build GRCh38
rs780586892 Chr.21: 25880676-25880678 on Build GRCh38
rs780567142 Chr.21: 25880712-25880712 on Build GRCh38
rs945440004 Chr.21: 25880801-25880801 on Build GRCh38
rs35871145 Chr.21: 25880676-25880676 on Build GRCh38
rs199738947 Chr.21: 25880516-25880516 on Build GRCh38
rs199885985 Chr.21: 25880873-25880873 on Build GRCh38
rs1059478 Chr.21: 25880612-25880612 on Build GRCh38
rs758913449 Chr.21: 25880607-25880607 on Build GRCh38
rs931066255 Chr.21: 25881032-25881032 on Build GRCh38
rs45541739 Chr.21: 25880703-25880703 on Build GRCh38
rs202057479 Chr.21: 25880878-25880878 on Build GRCh38
rs201961275 Chr.21: 25880993-25880993 on Build GRCh38
rs867304682 Chr.21: 25880803-25880803 on Build GRCh38
rs898264501 Chr.21: 25880763-25880763 on Build GRCh38
rs776885449 Chr.21: 25880967-25880967 on Build GRCh38
rs200544221 Chr.21: 25880611-25880611 on Build GRCh38
rs995244959 Chr.21: 25880772-25880772 on Build GRCh38
rs890648506 Chr.21: 25880539-25880539 on Build GRCh38
rs199913244 Chr.21: 25880609-25880609 on Build GRCh38
rs566047180 Chr.21: 25880847-25880847 on Build GRCh38
rs186409009 Chr.21: 25880953-25880953 on Build GRCh38
rs898132746 Chr.21: 25881013-25881013 on Build GRCh38
rs966712982 Chr.21: 25881013-25881013 on Build GRCh38
rs201885383 Chr.21: 25880853-25880853 on Build GRCh38
rs202108412 Chr.21: 25880655-25880655 on Build GRCh38
rs1015391498 Chr.21: 25880496-25880496 on Build GRCh38
rs3200120 Chr.21: 25880837-25880837 on Build GRCh38
rs45599935 Chr.21: 25880545-25880545 on Build GRCh38
rs200379862 Chr.21: 25880762-25880762 on Build GRCh38
rs945472589 Chr.21: 25880499-25880499 on Build GRCh38
rs200286647 Chr.21: 25880951-25880951 on Build GRCh38
rs200985813 Chr.21: 25880876-25880876 on Build GRCh38
rs989514534 Chr.21: 25880491-25880491 on Build GRCh38
rs11549660 Chr.21: 25881020-25881020 on Build GRCh38
rs572356515 Chr.21: 25881007-25881007 on Build GRCh38
rs201417585 Chr.21: 25880513-25880513 on Build GRCh38
rs1020893568 Chr.21: 25881012-25881012 on Build GRCh38
rs200089634 Chr.21: 25880892-25880892 on Build GRCh38
rs1039693147 Chr.21: 25880813-25880817 on Build GRCh38
rs199632072 Chr.21: 25880538-25880538 on Build GRCh38
rs201942871 Chr.21: 25880922-25880922 on Build GRCh38
rs199963586 Chr.21: 25880846-25880846 on Build GRCh38
rs200259364 Chr.21: 25880923-25880923 on Build GRCh38
rs939900852 Chr.21: 25880963-25880963 on Build GRCh38
rs200692470 Chr.21: 25880817-25880817 on Build GRCh38
rs200813390 Chr.21: 25880523-25880523 on Build GRCh38
rs200603813 Chr.21: 25880936-25880936 on Build GRCh38
rs376822208 Chr.21: 25880594-25880594 on Build GRCh38
rs911902366 Chr.21: 25880588-25880588 on Build GRCh38
rs199549432 Chr.21: 25880968-25880968 on Build GRCh38
rs981264791 Chr.21: 25880901-25880901 on Build GRCh38
rs533043229 Chr.21: 25880760-25880760 on Build GRCh38
rs936080642 Chr.21: 25880658-25880658 on Build GRCh38
rs906350862 Chr.21: 25880920-25880920 on Build GRCh38
rs200270218 Chr.21: 25880508-25880508 on Build GRCh38
rs201382660 Chr.21: 25880833-25880833 on Build GRCh38
rs899490790 Chr.21: 25880615-25880619 on Build GRCh38
rs912021116 Chr.21: 25881029-25881029 on Build GRCh38
rs201621005 Chr.21: 25880932-25880932 on Build GRCh38
rs201539056 Chr.21: 25880766-25880766 on Build GRCh38
rs199500588 Chr.21: 25880771-25880771 on Build GRCh38
rs146774213 Chr.21: 25880946-25880946 on Build GRCh38
rs924725706 Chr.21: 25880622-25880622 on Build GRCh38
rs199762023 Chr.21: 25880812-25880812 on Build GRCh38
rs199700901 Chr.21: 25880949-25880949 on Build GRCh38
rs201471700 Chr.21: 25880621-25880621 on Build GRCh38
rs112973660 Chr.21: 25880552-25880552 on Build GRCh38
rs962446225 Chr.21: 25880521-25880521 on Build GRCh38
rs1003975709 Chr.21: 25880717-25880717 on Build GRCh38
rs1059461 Chr.21: 25880733-25880733 on Build GRCh38
rs199902897 Chr.21: 25880657-25880657 on Build GRCh38
rs736479 Chr.21: 25880756-25880756 on Build GRCh38
rs970609897 Chr.21: 25880742-25880742 on Build GRCh38
rs542107566 Chr.21: 25880679-25880679 on Build GRCh38
rs200053802 Chr.21: 25880640-25880640 on Build GRCh38
rs200452592 Chr.21: 25880745-25880745 on Build GRCh38
rs912036895 Chr.21: 25880805-25880805 on Build GRCh38

View Full Product Details