Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20188544-20188726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB2
Entrez Gene ID
2706
Gene Name
gap junction protein beta 2
Gene Aliases
CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, HID, KID, NSRD1, PPK
UniGene
Hs.524894
Gene Chromosome Location
Chr.13: 20187463-20192975 on Build GRCh38
Transcripts
NM_004004.5 XM_011535049.2

SNP Detail

SNP ID Chromosome Location
rs951136665 Chr.13: 20188696-20188696 on Build GRCh38
rs1048745119 Chr.13: 20188589-20188589 on Build GRCh38
rs922232025 Chr.13: 20188665-20188665 on Build GRCh38
rs901550257 Chr.13: 20188652-20188652 on Build GRCh38
rs963997727 Chr.13: 20188611-20188611 on Build GRCh38
rs1018642379 Chr.13: 20188632-20188632 on Build GRCh38
rs990380831 Chr.13: 20188701-20188701 on Build GRCh38
rs747699704 Chr.13: 20188584-20188584 on Build GRCh38
rs552266792 Chr.13: 20188572-20188572 on Build GRCh38
rs371517351 Chr.13: 20188608-20188608 on Build GRCh38
rs957868023 Chr.13: 20188569-20188569 on Build GRCh38
rs771632395 Chr.13: 20188688-20188688 on Build GRCh38
rs886090162 Chr.13: 20188593-20188593 on Build GRCh38
rs537986752 Chr.13: 20188661-20188661 on Build GRCh38
rs976409791 Chr.13: 20188634-20188634 on Build GRCh38
rs996377279 Chr.13: 20188656-20188656 on Build GRCh38

View Full Product Details