Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62694157-62694702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
348498 c.428C>A p.S143Y Chr.11: 62694578-62694578 on Build GRCh38
6873301 c.387C>T p.T129T Chr.11: 62694619-62694619 on Build GRCh38
5015014 c.626G>A p.R209H Chr.11: 62694572-62694572 on Build GRCh38
5093614 c.313C>T p.P105S Chr.11: 62694693-62694693 on Build GRCh38
4199013 c.629C>T p.S210L Chr.11: 62694569-62694569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376639703 Chr.11: 62694542-62694542 on Build GRCh38
rs189367301 Chr.11: 62694307-62694307 on Build GRCh38
rs1052424900 Chr.11: 62694309-62694309 on Build GRCh38
rs772476337 Chr.11: 62694675-62694675 on Build GRCh38
rs921677845 Chr.11: 62694508-62694508 on Build GRCh38
rs771534001 Chr.11: 62694593-62694593 on Build GRCh38
rs184533368 Chr.11: 62694183-62694183 on Build GRCh38
rs777598096 Chr.11: 62694577-62694577 on Build GRCh38
rs150158560 Chr.11: 62694568-62694568 on Build GRCh38
rs1018384738 Chr.11: 62694413-62694413 on Build GRCh38
rs905672252 Chr.11: 62694396-62694396 on Build GRCh38
rs951977651 Chr.11: 62694444-62694444 on Build GRCh38
rs747968134 Chr.11: 62694570-62694570 on Build GRCh38
rs896554800 Chr.11: 62694336-62694336 on Build GRCh38
rs778486956 Chr.11: 62694569-62694569 on Build GRCh38
rs752392333 Chr.11: 62694672-62694672 on Build GRCh38
rs968763894 Chr.11: 62694390-62694390 on Build GRCh38
rs758029688 Chr.11: 62694673-62694673 on Build GRCh38
rs746973203 Chr.11: 62694578-62694578 on Build GRCh38
rs1013652887 Chr.11: 62694339-62694339 on Build GRCh38
rs947814112 Chr.11: 62694237-62694237 on Build GRCh38
rs983114637 Chr.11: 62694456-62694456 on Build GRCh38
rs759642646 Chr.11: 62694588-62694588 on Build GRCh38
rs571672404 Chr.11: 62694613-62694613 on Build GRCh38
rs751948915 Chr.11: 62694641-62694641 on Build GRCh38
rs753309219 Chr.11: 62694652-62694652 on Build GRCh38
rs752056113 Chr.11: 62694552-62694552 on Build GRCh38
rs144098421 Chr.11: 62694282-62694282 on Build GRCh38
rs587777607 Chr.11: 62694660-62694660 on Build GRCh38
rs892396839 Chr.11: 62694317-62694317 on Build GRCh38
rs745900103 Chr.11: 62694562-62694565 on Build GRCh38
rs965775441 Chr.11: 62694495-62694495 on Build GRCh38
rs199567191 Chr.11: 62694554-62694554 on Build GRCh38
rs774393283 Chr.11: 62694380-62694382 on Build GRCh38
rs369806785 Chr.11: 62694610-62694610 on Build GRCh38
rs765501396 Chr.11: 62694541-62694541 on Build GRCh38
rs398016308 Chr.11: 62694223-62694223 on Build GRCh38
rs886048442 Chr.11: 62694654-62694654 on Build GRCh38
rs1044875818 Chr.11: 62694241-62694241 on Build GRCh38
rs1048055867 Chr.11: 62694181-62694181 on Build GRCh38
rs906344464 Chr.11: 62694251-62694251 on Build GRCh38
rs544326129 Chr.11: 62694308-62694308 on Build GRCh38
rs761073584 Chr.11: 62694534-62694534 on Build GRCh38
rs137852970 Chr.11: 62694594-62694594 on Build GRCh38
rs374507554 Chr.11: 62694643-62694643 on Build GRCh38
rs930229044 Chr.11: 62694472-62694472 on Build GRCh38
rs1001777289 Chr.11: 62694401-62694401 on Build GRCh38
rs184922018 Chr.11: 62694457-62694457 on Build GRCh38
rs1006933787 Chr.11: 62694654-62694654 on Build GRCh38
rs768008962 Chr.11: 62694651-62694651 on Build GRCh38
rs965927987 Chr.11: 62694422-62694422 on Build GRCh38
rs904506245 Chr.11: 62694624-62694624 on Build GRCh38
rs11231190 Chr.11: 62694329-62694329 on Build GRCh38
rs1003400727 Chr.11: 62694306-62694306 on Build GRCh38
rs71056545 Chr.11: 62694196-62694196 on Build GRCh38
rs138532996 Chr.11: 62694574-62694574 on Build GRCh38
rs564013488 Chr.11: 62694407-62694407 on Build GRCh38
rs1024151382 Chr.11: 62694389-62694389 on Build GRCh38
rs760613992 Chr.11: 62694666-62694666 on Build GRCh38
rs779498581 Chr.11: 62694557-62694557 on Build GRCh38
rs537918937 Chr.11: 62694235-62694235 on Build GRCh38
rs763374609 Chr.11: 62694543-62694543 on Build GRCh38
rs764424992 Chr.11: 62694619-62694619 on Build GRCh38
rs1045195404 Chr.11: 62694554-62694556 on Build GRCh38
rs140208002 Chr.11: 62694583-62694583 on Build GRCh38
rs770894326 Chr.11: 62694582-62694582 on Build GRCh38
rs147314953 Chr.11: 62694483-62694483 on Build GRCh38
rs192698510 Chr.11: 62694437-62694437 on Build GRCh38
rs985938542 Chr.11: 62694402-62694402 on Build GRCh38
rs73483955 Chr.11: 62694504-62694504 on Build GRCh38
rs888155051 Chr.11: 62694195-62694195 on Build GRCh38
rs906321138 Chr.11: 62694563-62694563 on Build GRCh38
rs764526052 Chr.11: 62694547-62694547 on Build GRCh38
rs373161158 Chr.11: 62694527-62694527 on Build GRCh38
rs1021654614 Chr.11: 62694350-62694350 on Build GRCh38
rs1010202271 Chr.11: 62694338-62694338 on Build GRCh38

View Full Product Details