Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44793497-44794020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620916 c.1167C>T p.C389C Chr.19: 44793503-44793503 on Build GRCh38
6345643 c.1284+1G>T p.? Chr.19: 44793621-44793621 on Build GRCh38
7422023 c.1187G>T p.W396L Chr.19: 44793523-44793523 on Build GRCh38
3960196 c.1227T>A p.T409T Chr.19: 44793563-44793563 on Build GRCh38
5020654 c.1239A>G p.S413S Chr.19: 44793575-44793575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878873972 Chr.19: 44793722-44793722 on Build GRCh38
rs913923822 Chr.19: 44793709-44793709 on Build GRCh38
rs1043138391 Chr.19: 44793539-44793539 on Build GRCh38
rs913257227 Chr.19: 44793745-44793745 on Build GRCh38
rs199840214 Chr.19: 44793670-44793670 on Build GRCh38
rs994347051 Chr.19: 44793603-44793603 on Build GRCh38
rs866135435 Chr.19: 44793830-44793830 on Build GRCh38
rs869161737 Chr.19: 44793590-44793591 on Build GRCh38
rs3208856 Chr.19: 44793549-44793549 on Build GRCh38
rs867917078 Chr.19: 44793907-44793907 on Build GRCh38
rs890198244 Chr.19: 44793896-44793896 on Build GRCh38
rs1026506289 Chr.19: 44793871-44793871 on Build GRCh38
rs61750955 Chr.19: 44793575-44793575 on Build GRCh38
rs753854663 Chr.19: 44793584-44793584 on Build GRCh38
rs879001239 Chr.19: 44793712-44793712 on Build GRCh38
rs77920506 Chr.19: 44793591-44793591 on Build GRCh38
rs201433561 Chr.19: 44793578-44793578 on Build GRCh38
rs375842565 Chr.19: 44793657-44793657 on Build GRCh38
rs768506085 Chr.19: 44793575-44793576 on Build GRCh38
rs901313209 Chr.19: 44793901-44793901 on Build GRCh38
rs988722908 Chr.19: 44793724-44793724 on Build GRCh38
rs772388756 Chr.19: 44793558-44793558 on Build GRCh38
rs571579481 Chr.19: 44793969-44793969 on Build GRCh38
rs374717459 Chr.19: 44793635-44793635 on Build GRCh38
rs762183942 Chr.19: 44793588-44793588 on Build GRCh38
rs754849774 Chr.19: 44793595-44793595 on Build GRCh38
rs112128460 Chr.19: 44793530-44793530 on Build GRCh38
rs769396781 Chr.19: 44793518-44793518 on Build GRCh38
rs531857703 Chr.19: 44793822-44793822 on Build GRCh38
rs1047296929 Chr.19: 44793955-44793955 on Build GRCh38
rs993264052 Chr.19: 44793814-44793814 on Build GRCh38
rs749036591 Chr.19: 44793531-44793531 on Build GRCh38
rs369869473 Chr.19: 44793744-44793745 on Build GRCh38
rs945591284 Chr.19: 44793714-44793714 on Build GRCh38
rs17852620 Chr.19: 44793573-44793573 on Build GRCh38
rs113457155 Chr.19: 44793887-44793887 on Build GRCh38
rs551783911 Chr.19: 44793905-44793905 on Build GRCh38
rs118085256 Chr.19: 44793637-44793637 on Build GRCh38
rs949439657 Chr.19: 44793764-44793765 on Build GRCh38
rs370803830 Chr.19: 44793826-44793826 on Build GRCh38
rs748765725 Chr.19: 44793643-44793643 on Build GRCh38
rs975600354 Chr.19: 44793979-44793981 on Build GRCh38
rs756001540 Chr.19: 44793636-44793636 on Build GRCh38
rs539714533 Chr.19: 44793985-44793985 on Build GRCh38
rs765569181 Chr.19: 44793591-44793591 on Build GRCh38
rs796196600 Chr.19: 44793830-44793831 on Build GRCh38
rs750604085 Chr.19: 44793593-44793593 on Build GRCh38
rs928424019 Chr.19: 44793676-44793676 on Build GRCh38
rs538290840 Chr.19: 44793764-44793764 on Build GRCh38
rs947394585 Chr.19: 44793770-44793770 on Build GRCh38
rs560728785 Chr.19: 44793617-44793617 on Build GRCh38
rs368494441 Chr.19: 44793552-44793552 on Build GRCh38
rs753086782 Chr.19: 44793837-44793837 on Build GRCh38
rs770365728 Chr.19: 44793650-44793650 on Build GRCh38
rs762865111 Chr.19: 44793531-44793532 on Build GRCh38
rs898965307 Chr.19: 44793792-44793792 on Build GRCh38
rs781113299 Chr.19: 44793597-44793597 on Build GRCh38
rs1009013289 Chr.19: 44793939-44793939 on Build GRCh38
rs989655688 Chr.19: 44793711-44793711 on Build GRCh38
rs778570821 Chr.19: 44793651-44793651 on Build GRCh38
rs756858266 Chr.19: 44793685-44793686 on Build GRCh38
rs950413191 Chr.19: 44793614-44793614 on Build GRCh38
rs764271266 Chr.19: 44793583-44793583 on Build GRCh38
rs1041206889 Chr.19: 44793883-44793883 on Build GRCh38
rs868456325 Chr.19: 44793683-44793683 on Build GRCh38
rs1025942028 Chr.19: 44793612-44793612 on Build GRCh38
rs777117772 Chr.19: 44793529-44793529 on Build GRCh38
rs886134180 Chr.19: 44793961-44793961 on Build GRCh38
rs1053116620 Chr.19: 44793516-44793516 on Build GRCh38
rs773939139 Chr.19: 44793551-44793551 on Build GRCh38
rs1025992203 Chr.19: 44793581-44793581 on Build GRCh38
rs1043555930 Chr.19: 44793774-44793774 on Build GRCh38
rs905143355 Chr.19: 44793773-44793773 on Build GRCh38
rs772470272 Chr.19: 44793658-44793658 on Build GRCh38
rs66944506 Chr.19: 44793589-44793590 on Build GRCh38
rs938583306 Chr.19: 44793691-44793691 on Build GRCh38
rs569345016 Chr.19: 44793783-44793783 on Build GRCh38
rs778143095 Chr.19: 44793946-44793946 on Build GRCh38
rs200751399 Chr.19: 44793717-44793717 on Build GRCh38

View Full Product Details