Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164391.1 | NP_001157863.1 | 11 - 12 | 1204 | 89 |
NM_001164393.1 | NP_001157865.1 | 8 - 9 | 1003 | 89 | |
NM_007358.3 | NP_031384.1 | 10 - 11 | 1284 | 89 | |
XM_011541016.1 | XP_011539318.1 | 10 - 11 | 1240 | 89 | |
XM_011541019.1 | XP_011539321.1 | 10 - 11 | 1032 | 89 | |
XM_017000676.1 | XP_016856165.1 | 10 - 11 | 1139 | 89 | |
XM_017000679.1 | XP_016856168.1 | 9 - 10 | 1116 | 89 | |
GenBank mRNA | AF072814.1 | - | 9 - 10 | 982 | 89 |
AF086908.1 | - | 9 - 10 | 1085 | 89 | |
AJ010014.1 | - | 10 - 11 | 1235 | 89 | |
AK290318.1 | - | 10 - 11 | 1236 | 89 | |
AK302776.1 | - | 11 - 12 | 1143 | 89 | |
AK302900.1 | - | 8 - 9 | 955 | 89 | |
BC001897.1 | - | 10 - 11 | 1203 | 89 |