Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18121268-18121804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC1
Entrez Gene ID
378884
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
Gene Aliases
EPM2A, EPM2B, MALIN, bA204B7.2
UniGene
Hs.348351
Gene Chromosome Location
Chr.6: 18120487-18122620 on Build GRCh38
Transcripts
NM_198586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
450955 c.1139T>C p.L380P Chr.6: 18121468-18121468 on Build GRCh38
6455892 c.869G>T p.C290F Chr.6: 18121738-18121738 on Build GRCh38
450956 c.977C>T p.T326I Chr.6: 18121630-18121630 on Build GRCh38
2856205 c.904C>T p.Q302* Chr.6: 18121703-18121703 on Build GRCh38
1621242 c.1165G>A p.V389I Chr.6: 18121442-18121442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11966789 Chr.6: 18121364-18121364 on Build GRCh38
rs587780400 Chr.6: 18121634-18121634 on Build GRCh38
rs775777721 Chr.6: 18121642-18121642 on Build GRCh38
rs774397860 Chr.6: 18121657-18121657 on Build GRCh38
rs757858146 Chr.6: 18121702-18121702 on Build GRCh38
rs998873745 Chr.6: 18121350-18121350 on Build GRCh38
rs765443391 Chr.6: 18121535-18121535 on Build GRCh38
rs750770846 Chr.6: 18121421-18121422 on Build GRCh38
rs865887763 Chr.6: 18121480-18121480 on Build GRCh38
rs201855901 Chr.6: 18121742-18121742 on Build GRCh38
rs746839446 Chr.6: 18121627-18121627 on Build GRCh38
rs73379121 Chr.6: 18121334-18121334 on Build GRCh38
rs200201752 Chr.6: 18121465-18121465 on Build GRCh38
rs780904929 Chr.6: 18121728-18121728 on Build GRCh38
rs1029613778 Chr.6: 18121329-18121329 on Build GRCh38
rs377395116 Chr.6: 18121554-18121554 on Build GRCh38
rs781484428 Chr.6: 18121459-18121459 on Build GRCh38
rs377500729 Chr.6: 18121381-18121381 on Build GRCh38
rs756437109 Chr.6: 18121759-18121759 on Build GRCh38
rs751956775 Chr.6: 18121547-18121547 on Build GRCh38
rs376530552 Chr.6: 18121592-18121592 on Build GRCh38
rs981548932 Chr.6: 18121410-18121410 on Build GRCh38
rs764407367 Chr.6: 18121546-18121546 on Build GRCh38
rs756591398 Chr.6: 18121581-18121581 on Build GRCh38
rs747655441 Chr.6: 18121590-18121590 on Build GRCh38
rs142941035 Chr.6: 18121638-18121638 on Build GRCh38
rs766225081 Chr.6: 18121423-18121423 on Build GRCh38
rs970281032 Chr.6: 18121402-18121402 on Build GRCh38
rs765576968 Chr.6: 18121433-18121433 on Build GRCh38
rs1032004459 Chr.6: 18121738-18121738 on Build GRCh38
rs554356127 Chr.6: 18121751-18121751 on Build GRCh38
rs78324544 Chr.6: 18121516-18121516 on Build GRCh38
rs137852859 Chr.6: 18121684-18121684 on Build GRCh38
rs527299943 Chr.6: 18121674-18121674 on Build GRCh38
rs556216487 Chr.6: 18121776-18121776 on Build GRCh38
rs370044232 Chr.6: 18121517-18121517 on Build GRCh38
rs773945358 Chr.6: 18121401-18121401 on Build GRCh38
rs757847339 Chr.6: 18121454-18121454 on Build GRCh38
rs562100142 Chr.6: 18121441-18121441 on Build GRCh38
rs886061252 Chr.6: 18121358-18121358 on Build GRCh38
rs760726977 Chr.6: 18121422-18121422 on Build GRCh38
rs763060322 Chr.6: 18121644-18121644 on Build GRCh38
rs745652384 Chr.6: 18121732-18121732 on Build GRCh38
rs752220824 Chr.6: 18121694-18121694 on Build GRCh38
rs778329369 Chr.6: 18121440-18121440 on Build GRCh38
rs1045381882 Chr.6: 18121500-18121500 on Build GRCh38
rs994146351 Chr.6: 18121688-18121688 on Build GRCh38
rs750935925 Chr.6: 18121580-18121580 on Build GRCh38
rs1031632114 Chr.6: 18121358-18121358 on Build GRCh38
rs767777371 Chr.6: 18121769-18121769 on Build GRCh38
rs747848818 Chr.6: 18121482-18121482 on Build GRCh38
rs761628913 Chr.6: 18121405-18121405 on Build GRCh38
rs1029911244 Chr.6: 18121377-18121377 on Build GRCh38
rs764389649 Chr.6: 18121647-18121647 on Build GRCh38
rs768659412 Chr.6: 18121743-18121743 on Build GRCh38
rs375475285 Chr.6: 18121752-18121752 on Build GRCh38
rs771702699 Chr.6: 18121487-18121487 on Build GRCh38
rs758659117 Chr.6: 18121436-18121436 on Build GRCh38
rs945725973 Chr.6: 18121483-18121483 on Build GRCh38
rs587776543 Chr.6: 18121615-18121615 on Build GRCh38
rs778456340 Chr.6: 18121586-18121586 on Build GRCh38
rs773130728 Chr.6: 18121489-18121489 on Build GRCh38
rs750788641 Chr.6: 18121680-18121680 on Build GRCh38
rs545172272 Chr.6: 18121597-18121597 on Build GRCh38
rs752924111 Chr.6: 18121435-18121435 on Build GRCh38
rs746676778 Chr.6: 18121713-18121713 on Build GRCh38
rs911410366 Chr.6: 18121413-18121413 on Build GRCh38
rs777509826 Chr.6: 18121711-18121711 on Build GRCh38
rs758056041 Chr.6: 18121600-18121600 on Build GRCh38
rs760322295 Chr.6: 18121515-18121515 on Build GRCh38
rs772972107 Chr.6: 18121409-18121409 on Build GRCh38
rs748854501 Chr.6: 18121462-18121462 on Build GRCh38
rs776454075 Chr.6: 18121637-18121637 on Build GRCh38
rs770587249 Chr.6: 18121630-18121630 on Build GRCh38
rs989899382 Chr.6: 18121468-18121468 on Build GRCh38
rs372993582 Chr.6: 18121531-18121531 on Build GRCh38
rs772460190 Chr.6: 18121398-18121398 on Build GRCh38
rs140066702 Chr.6: 18121770-18121770 on Build GRCh38
rs766807637 Chr.6: 18121689-18121689 on Build GRCh38
rs756909945 Chr.6: 18121727-18121727 on Build GRCh38
rs979143914 Chr.6: 18121678-18121678 on Build GRCh38
rs148553723 Chr.6: 18121617-18121617 on Build GRCh38
rs753959228 Chr.6: 18121424-18121424 on Build GRCh38
rs531663760 Chr.6: 18121417-18121417 on Build GRCh38
rs144863228 Chr.6: 18121543-18121543 on Build GRCh38
rs759473681 Chr.6: 18121425-18121425 on Build GRCh38
rs752799460 Chr.6: 18121542-18121542 on Build GRCh38
rs778364234 Chr.6: 18121475-18121475 on Build GRCh38
rs543997890 Chr.6: 18121437-18121437 on Build GRCh38
rs762064606 Chr.6: 18121664-18121664 on Build GRCh38
rs756683363 Chr.6: 18121685-18121685 on Build GRCh38
rs541460675 Chr.6: 18121442-18121442 on Build GRCh38
rs193252025 Chr.6: 18121407-18121407 on Build GRCh38
rs781739043 Chr.6: 18121573-18121573 on Build GRCh38
rs748741304 Chr.6: 18121384-18121384 on Build GRCh38
rs762001530 Chr.6: 18121764-18121764 on Build GRCh38
rs996662904 Chr.6: 18121304-18121304 on Build GRCh38
rs1029669440 Chr.6: 18121700-18121700 on Build GRCh38
rs200214191 Chr.6: 18121733-18121733 on Build GRCh38

View Full Product Details