Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FHOD1 | NM_001318202.1 | NP_001305131.1 | ||
NM_013241.2 | NP_037373.2 | |||
XM_006721180.1 | XP_006721243.1 | |||
XM_011523043.2 | XP_011521345.1 | |||
XM_011523044.1 | XP_011521346.1 | |||
XM_011523045.2 | XP_011521347.1 | |||
AB041046.1 | BAD06250.1 | |||
AB209584.1 | BAD92821.1 | |||
AF113615.1 | AAD39906.1 | |||
AK025992.1 | ||||
AK301157.1 | ||||
AY192154.1 | AAO38757.1 | |||
BC033084.1 | AAH33084.1 | |||
KJ658417.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr16:67153308 - 68071571 on Build GRCh38 | Gain | CTRL LCAT NRN1L EXOC3L1 CTCF LOC107984813 RANBP10 TPPP3 DPEP3 THAP11 SLC9A5 HSD11B2 LOC100505942 FAM65A FBXL8 TSNAXIP1 EDC4 LOC100131303 DPEP2 FHOD1 ZDHHC1 CARMIL2 PARD6A KCTD19 ATP6V0D1 ELMO3 NOL3 DUS2 HSF4 MIR328 NUTF2 E2F4 KIAA0895L LRRC29 TMEM208 PLEKHG4 LRRC36 LOC105369155 AGRP PSMB10 GFOD2 SLC12A4 CENPT C16orf86 ACD TRADD DDX28 PSKH1 ENKD1 |
nsv827707 | Chr16:67148355 - 67289296 on Build GRCh38 | Loss | LOC105369155 C16orf70 FBXL8 ELMO3 B3GNT9 EXOC3L1 NOL3 FHOD1 HSF4 MIR328 TRADD E2F4 KIAA0895L LRRC29 TMEM208 PLEKHG4 SLC9A5 |
nsv952044 | Chr16:67141198 - 67244397 on Build GRCh38 | Deletion | LOC105369155 C16orf70 FBXL8 ELMO3 B3GNT9 EXOC3L1 NOL3 FHOD1 HSF4 MIR328 TRADD E2F4 KIAA0895L LRRC29 TMEM208 |
nsv457513 | Chr16:67181417 - 67282697 on Build GRCh38 | Loss | LOC105369155 EXOC3L1 FHOD1 MIR328 E2F4 KIAA0895L LRRC29 ELMO3 TMEM208 PLEKHG4 SLC9A5 |
nsv524492 | Chr16:67174030 - 67294550 on Build GRCh38 | Loss | LOC105369155 KCTD19 ELMO3 EXOC3L1 NOL3 FHOD1 MIR328 E2F4 KIAA0895L LRRC29 TMEM208 PLEKHG4 SLC9A5 |
nsv524363 | Chr16:67136529 - 68071571 on Build GRCh38 | Loss | CTRL LCAT NRN1L B3GNT9 EXOC3L1 CTCF LOC107984813 RANBP10 TPPP3 DPEP3 THAP11 SLC9A5 HSD11B2 C16orf70 LOC100505942 FAM65A FBXL8 TSNAXIP1 EDC4 LOC100131303 DPEP2 FHOD1 ZDHHC1 CARMIL2 PARD6A KCTD19 ATP6V0D1 ELMO3 NOL3 DUS2 HSF4 MIR328 NUTF2 E2F4 KIAA0895L LRRC29 TMEM208 PLEKHG4 LRRC36 LOC105369155 AGRP PSMB10 GFOD2 SLC12A4 CENPT C16orf86 ACD TRADD DDX28 PSKH1 ENKD1 |