Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001132.1 | NP_001001132.1 | 25 - 26 | 3450 | 71 |
NM_003024.2 | NP_003015.2 | 25 - 26 | 3450 | 71 | |
XM_005261026.2 | XP_005261083.1 | 24 - 25 | 3513 | 71 | |
XM_005261027.1 | XP_005261084.1 | 24 - 25 | 3513 | 71 | |
XM_011529691.1 | XP_011527993.1 | 24 - 25 | 3402 | 71 | |
XM_017028428.1 | XP_016883917.1 | 26 - 27 | 3448 | 71 | |
XM_017028429.1 | XP_016883918.1 | 25 - 26 | 3417 | 71 | |
XM_017028430.1 | XP_016883919.1 | 24 - 25 | 3402 | 71 | |
XM_017028431.1 | XP_016883920.1 | 25 - 26 | 3528 | 71 | |
XM_017028432.1 | XP_016883921.1 | 24 - 25 | 3513 | 71 | |
XM_017028437.1 | XP_016883926.1 | 20 - 21 | 2868 | 71 | |
XM_017028438.1 | XP_016883927.1 | 25 - 26 | 3417 | 71 | |
GenBank mRNA | AF064243.1 | - | 25 - 26 | 3288 | 71 |
AF064244.1 | - | 25 - 26 | 3288 | 71 | |
AF114487.1 | - | 25 - 26 | 3450 | 71 | |
AF114488.1 | - | 25 - 26 | 3450 | 71 | |
AK300274.1 | - | 24 - 25 | 3410 | 71 | |
AY340592.1 | - | 3 - 4 | 339 | 71 | |
BC116185.1 | - | 24 - 25 | 3246 | 71 | |
BC116186.2 | - | 25 - 26 | 3261 | 71 | |
DQ340865.1 | - | 23 - 24 | 3163 | 71 | |
DQ679756.1 | - | 25 - 26 | 3141 | 71 | |
DQ679757.1 | - | 24 - 25 | 3252 | 71 | |
EU107386.1 | - | 25 - 26 | 3156 | 71 | |
EU120733.1 | - | 23 - 24 | 3168 | 71 | |
EU120734.1 | - | 23 - 24 | 3168 | 71 | |
EU191901.1 | - | 23 - 24 | 3091 | 71 |