Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1995657-1996161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H19
Entrez Gene ID
283120
Gene Name
H19, imprinted maternally expressed transcript (non-protein coding)
Gene Aliases
ASM, ASM1, BWS, D11S813E, LINC00008, NCRNA00008, WT2
UniGene
Hs.533566
Gene Chromosome Location
Chr.11: 1995176-2001466 on Build GRCh38
Transcripts
NR_002196.2 NR_131223.1 NR_131224.1 NM_001293171.2

Gene Symbol
HOTS
Entrez Gene ID
103344718
Gene Name
H19 opposite tumor suppressor
Gene Aliases
-
UniGene
Hs.533566
Gene Chromosome Location
Chr.11: 1992408-1996294 on Build GRCh38
Transcripts
NR_002196.2 NR_131223.1 NR_131224.1 NM_001293171.2

SNP Detail

SNP ID Chromosome Location
rs185407010 Chr.11: 1995797-1995797 on Build GRCh38
rs757761252 Chr.11: 1995810-1995810 on Build GRCh38
rs572306720 Chr.11: 1995854-1995854 on Build GRCh38
rs778011121 Chr.11: 1996086-1996086 on Build GRCh38
rs898526401 Chr.11: 1996046-1996046 on Build GRCh38
rs545220745 Chr.11: 1996127-1996127 on Build GRCh38
rs868152285 Chr.11: 1996083-1996083 on Build GRCh38
rs771161376 Chr.11: 1995733-1995733 on Build GRCh38
rs762931682 Chr.11: 1995747-1995747 on Build GRCh38
rs762498693 Chr.11: 1995977-1995977 on Build GRCh38
rs755785499 Chr.11: 1995718-1995718 on Build GRCh38
rs111714200 Chr.11: 1996003-1996003 on Build GRCh38
rs769486921 Chr.11: 1995960-1995960 on Build GRCh38
rs1024588105 Chr.11: 1996097-1996097 on Build GRCh38
rs774087209 Chr.11: 1995766-1995766 on Build GRCh38
rs747526239 Chr.11: 1995981-1995981 on Build GRCh38
rs769028996 Chr.11: 1995694-1995699 on Build GRCh38
rs763137072 Chr.11: 1995707-1995707 on Build GRCh38
rs550180086 Chr.11: 1995681-1995681 on Build GRCh38
rs763704036 Chr.11: 1995979-1995979 on Build GRCh38
rs776466438 Chr.11: 1995938-1995938 on Build GRCh38
rs754360673 Chr.11: 1996053-1996053 on Build GRCh38
rs758599358 Chr.11: 1996073-1996073 on Build GRCh38
rs750034401 Chr.11: 1995716-1995716 on Build GRCh38
rs772197020 Chr.11: 1995917-1995917 on Build GRCh38
rs112056845 Chr.11: 1995871-1995871 on Build GRCh38
rs749141470 Chr.11: 1996118-1996118 on Build GRCh38
rs544222336 Chr.11: 1996051-1996051 on Build GRCh38
rs773972518 Chr.11: 1996050-1996050 on Build GRCh38
rs750983016 Chr.11: 1995882-1995882 on Build GRCh38
rs11542716 Chr.11: 1995926-1995926 on Build GRCh38
rs765523428 Chr.11: 1996067-1996067 on Build GRCh38
rs760719856 Chr.11: 1996007-1996007 on Build GRCh38
rs2839701 Chr.11: 1995880-1995880 on Build GRCh38
rs768791984 Chr.11: 1995752-1995752 on Build GRCh38
rs779181816 Chr.11: 1996034-1996034 on Build GRCh38
rs369668916 Chr.11: 1995892-1995892 on Build GRCh38
rs770113730 Chr.11: 1995744-1995744 on Build GRCh38
rs113390616 Chr.11: 1996078-1996078 on Build GRCh38
rs748345774 Chr.11: 1995860-1995863 on Build GRCh38
rs2839702 Chr.11: 1995787-1995787 on Build GRCh38
rs781775699 Chr.11: 1995811-1995811 on Build GRCh38
rs766168463 Chr.11: 1995856-1995856 on Build GRCh38
rs775693856 Chr.11: 1995699-1995699 on Build GRCh38
rs754196445 Chr.11: 1996029-1996029 on Build GRCh38
rs761837808 Chr.11: 1995773-1995773 on Build GRCh38
rs752873808 Chr.11: 1996035-1996035 on Build GRCh38
rs371414038 Chr.11: 1995805-1995805 on Build GRCh38
rs752868692 Chr.11: 1996072-1996072 on Build GRCh38
rs781130471 Chr.11: 1996109-1996109 on Build GRCh38
rs771966034 Chr.11: 1995844-1995844 on Build GRCh38
rs217727 Chr.11: 1995678-1995678 on Build GRCh38
rs199912364 Chr.11: 1996047-1996047 on Build GRCh38
rs753545443 Chr.11: 1995788-1995788 on Build GRCh38
rs759289987 Chr.11: 1995959-1995959 on Build GRCh38
rs751635496 Chr.11: 1996108-1996108 on Build GRCh38
rs778086830 Chr.11: 1995730-1995730 on Build GRCh38
rs774508787 Chr.11: 1995834-1995834 on Build GRCh38
rs578029526 Chr.11: 1996115-1996115 on Build GRCh38
rs935650049 Chr.11: 1995973-1995973 on Build GRCh38
rs929296565 Chr.11: 1995948-1995959 on Build GRCh38
rs770666062 Chr.11: 1995937-1995937 on Build GRCh38
rs199553415 Chr.11: 1995884-1995885 on Build GRCh38
rs374736833 Chr.11: 1995864-1995864 on Build GRCh38
rs747556950 Chr.11: 1995732-1995732 on Build GRCh38
rs767247053 Chr.11: 1995782-1995782 on Build GRCh38
rs745666058 Chr.11: 1996045-1996045 on Build GRCh38
rs771896100 Chr.11: 1996117-1996117 on Build GRCh38
rs369974003 Chr.11: 1995879-1995879 on Build GRCh38
rs755363476 Chr.11: 1996032-1996032 on Build GRCh38
rs751574748 Chr.11: 1995710-1995710 on Build GRCh38
rs10840159 Chr.11: 1995694-1995694 on Build GRCh38
rs772270729 Chr.11: 1995931-1995932 on Build GRCh38
rs756608025 Chr.11: 1995815-1995815 on Build GRCh38
rs754582297 Chr.11: 1995729-1995729 on Build GRCh38
rs777854336 Chr.11: 1996039-1996039 on Build GRCh38
rs768229301 Chr.11: 1996048-1996048 on Build GRCh38
rs747141756 Chr.11: 1996040-1996040 on Build GRCh38
rs754381130 Chr.11: 1995795-1995795 on Build GRCh38
rs375233458 Chr.11: 1995886-1995886 on Build GRCh38
rs775924831 Chr.11: 1995745-1995745 on Build GRCh38
rs780368864 Chr.11: 1995817-1995817 on Build GRCh38
rs781707071 Chr.11: 1995734-1995734 on Build GRCh38
rs373536357 Chr.11: 1995896-1995896 on Build GRCh38
rs775067281 Chr.11: 1995970-1995970 on Build GRCh38
rs751962180 Chr.11: 1995808-1995808 on Build GRCh38
rs149202370 Chr.11: 1996132-1996132 on Build GRCh38
rs866347619 Chr.11: 1996057-1996057 on Build GRCh38
rs760447421 Chr.11: 1995855-1995855 on Build GRCh38
rs746288770 Chr.11: 1995739-1995739 on Build GRCh38
rs774584953 Chr.11: 1995749-1995750 on Build GRCh38
rs771341828 Chr.11: 1995687-1995687 on Build GRCh38
rs779043642 Chr.11: 1995898-1995898 on Build GRCh38
rs764847370 Chr.11: 1995866-1995866 on Build GRCh38
rs749526897 Chr.11: 1995826-1995826 on Build GRCh38
rs889870495 Chr.11: 1995746-1995746 on Build GRCh38
rs12577355 Chr.11: 1995992-1995992 on Build GRCh38
rs745800394 Chr.11: 1996111-1996111 on Build GRCh38
rs553988547 Chr.11: 1995836-1995836 on Build GRCh38
rs769945357 Chr.11: 1995695-1995695 on Build GRCh38
rs746982390 Chr.11: 1995929-1995929 on Build GRCh38
rs761454175 Chr.11: 1995711-1995711 on Build GRCh38
rs369312603 Chr.11: 1996133-1996133 on Build GRCh38
rs867483975 Chr.11: 1996056-1996056 on Build GRCh38
rs575848160 Chr.11: 1995863-1995863 on Build GRCh38
rs747646425 Chr.11: 1996128-1996128 on Build GRCh38
rs753314512 Chr.11: 1995726-1995726 on Build GRCh38
rs768575700 Chr.11: 1995831-1995831 on Build GRCh38
rs772968023 Chr.11: 1995784-1995784 on Build GRCh38
rs771019983 Chr.11: 1996085-1996087 on Build GRCh38
rs562623966 Chr.11: 1996062-1996062 on Build GRCh38
rs111282089 Chr.11: 1995798-1995798 on Build GRCh38
rs773460981 Chr.11: 1995996-1995996 on Build GRCh38
rs368244047 Chr.11: 1995677-1995677 on Build GRCh38
rs780593904 Chr.11: 1995888-1995888 on Build GRCh38
rs113259797 Chr.11: 1996001-1996001 on Build GRCh38
rs980299693 Chr.11: 1996126-1996126 on Build GRCh38
rs761282976 Chr.11: 1996016-1996016 on Build GRCh38
rs113046940 Chr.11: 1995870-1995870 on Build GRCh38
rs748253962 Chr.11: 1995908-1995908 on Build GRCh38
rs747328776 Chr.11: 1995685-1995685 on Build GRCh38
rs768388545 Chr.11: 1996122-1996122 on Build GRCh38
rs776928091 Chr.11: 1995692-1995692 on Build GRCh38
rs770823815 Chr.11: 1996041-1996041 on Build GRCh38
rs781062377 Chr.11: 1996042-1996042 on Build GRCh38
rs766840094 Chr.11: 1996027-1996027 on Build GRCh38
rs779730310 Chr.11: 1995719-1995719 on Build GRCh38

View Full Product Details