Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 22136286-22136798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
UniGene
Hs.712707
Gene Chromosome Location
Chr.6: 21666444-22194400 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

Gene Symbol
NBAT1
Entrez Gene ID
729177
Gene Name
neuroblastoma associated transcript 1
Gene Aliases
CASC14, NBAT-1
UniGene
Hs.730673
Gene Chromosome Location
Chr.6: 22134602-22147193 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

SNP Detail

SNP ID Chromosome Location
rs544307979 Chr.6: 22136665-22136665 on Build GRCh38
rs575510162 Chr.6: 22136412-22136412 on Build GRCh38
rs995804416 Chr.6: 22136601-22136601 on Build GRCh38
rs967155231 Chr.6: 22136401-22136401 on Build GRCh38
rs78638042 Chr.6: 22136610-22136610 on Build GRCh38
rs969546952 Chr.6: 22136476-22136476 on Build GRCh38
rs759352069 Chr.6: 22136717-22136717 on Build GRCh38
rs917958945 Chr.6: 22136430-22136430 on Build GRCh38
rs751673144 Chr.6: 22136599-22136599 on Build GRCh38
rs1039851846 Chr.6: 22136551-22136551 on Build GRCh38
rs141405065 Chr.6: 22136471-22136471 on Build GRCh38
rs981266836 Chr.6: 22136438-22136438 on Build GRCh38
rs1029411495 Chr.6: 22136387-22136387 on Build GRCh38
rs552307078 Chr.6: 22136331-22136331 on Build GRCh38
rs868753520 Chr.6: 22136516-22136516 on Build GRCh38
rs937298205 Chr.6: 22136456-22136456 on Build GRCh38
rs79555544 Chr.6: 22136535-22136535 on Build GRCh38
rs1051397227 Chr.6: 22136607-22136607 on Build GRCh38
rs941367978 Chr.6: 22136711-22136711 on Build GRCh38
rs977056193 Chr.6: 22136709-22136709 on Build GRCh38
rs757700120 Chr.6: 22136603-22136603 on Build GRCh38
rs563181686 Chr.6: 22136705-22136705 on Build GRCh38
rs775163866 Chr.6: 22136338-22136338 on Build GRCh38
rs1027013245 Chr.6: 22136394-22136394 on Build GRCh38
rs186644563 Chr.6: 22136634-22136634 on Build GRCh38
rs375119822 Chr.6: 22136433-22136433 on Build GRCh38
rs572996843 Chr.6: 22136646-22136646 on Build GRCh38
rs6921113 Chr.6: 22136650-22136650 on Build GRCh38
rs570420400 Chr.6: 22136411-22136411 on Build GRCh38
rs184847896 Chr.6: 22136337-22136337 on Build GRCh38
rs1054766838 Chr.6: 22136459-22136459 on Build GRCh38
rs565055081 Chr.6: 22136652-22136652 on Build GRCh38
rs928161286 Chr.6: 22136704-22136704 on Build GRCh38
rs369597483 Chr.6: 22136524-22136524 on Build GRCh38
rs6900792 Chr.6: 22136709-22136709 on Build GRCh38
rs746635703 Chr.6: 22136689-22136689 on Build GRCh38
rs115214299 Chr.6: 22136708-22136708 on Build GRCh38
rs1004133660 Chr.6: 22136613-22136613 on Build GRCh38
rs768434386 Chr.6: 22136314-22136314 on Build GRCh38
rs982729555 Chr.6: 22136483-22136483 on Build GRCh38
rs989722631 Chr.6: 22136437-22136437 on Build GRCh38
rs927219637 Chr.6: 22136450-22136450 on Build GRCh38
rs535622429 Chr.6: 22136574-22136574 on Build GRCh38
rs78938490 Chr.6: 22136654-22136654 on Build GRCh38
rs753947089 Chr.6: 22136609-22136609 on Build GRCh38
rs1025025343 Chr.6: 22136630-22136630 on Build GRCh38
rs545291214 Chr.6: 22136679-22136680 on Build GRCh38
rs1039584607 Chr.6: 22136562-22136562 on Build GRCh38
rs1040142358 Chr.6: 22136729-22136729 on Build GRCh38
rs998429361 Chr.6: 22136382-22136382 on Build GRCh38
rs192440368 Chr.6: 22136314-22136314 on Build GRCh38
rs941378338 Chr.6: 22136548-22136548 on Build GRCh38
rs982291150 Chr.6: 22136701-22136701 on Build GRCh38
rs374820301 Chr.6: 22136488-22136488 on Build GRCh38

View Full Product Details