Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35657489-35657994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMRP
Entrez Gene ID
6023
Gene Name
RNA component of mitochondrial RNA processing endoribonuclease
Gene Aliases
CHH, NME1, RMRPR, RRP2
UniGene
Hs.587502
Gene Chromosome Location
Chr.9: 35657751-35658018 on Build GRCh38
Transcripts
NR_003051.3

SNP Detail

SNP ID Chromosome Location
rs772825736 Chr.9: 35657906-35657906 on Build GRCh38
rs938089991 Chr.9: 35657894-35657894 on Build GRCh38
rs551450545 Chr.9: 35657776-35657776 on Build GRCh38
rs746593421 Chr.9: 35657665-35657665 on Build GRCh38
rs571495691 Chr.9: 35657643-35657643 on Build GRCh38
rs533294975 Chr.9: 35657757-35657757 on Build GRCh38
rs756128568 Chr.9: 35657939-35657939 on Build GRCh38
rs958947439 Chr.9: 35657527-35657527 on Build GRCh38
rs944727411 Chr.9: 35657669-35657669 on Build GRCh38
rs769854200 Chr.9: 35657768-35657768 on Build GRCh38
rs544069410 Chr.9: 35657957-35657957 on Build GRCh38
rs933259658 Chr.9: 35657830-35657830 on Build GRCh38
rs775067778 Chr.9: 35657747-35657747 on Build GRCh38
rs71521283 Chr.9: 35657744-35657744 on Build GRCh38
rs1033305693 Chr.9: 35657742-35657742 on Build GRCh38
rs572399988 Chr.9: 35657751-35657751 on Build GRCh38
rs749667892 Chr.9: 35657780-35657780 on Build GRCh38
rs555390900 Chr.9: 35657750-35657750 on Build GRCh38
rs528442230 Chr.9: 35657628-35657628 on Build GRCh38
rs940224067 Chr.9: 35657883-35657883 on Build GRCh38
rs754810221 Chr.9: 35657859-35657859 on Build GRCh38
rs550203343 Chr.9: 35657856-35657856 on Build GRCh38
rs991322091 Chr.9: 35657934-35657934 on Build GRCh38
rs150183436 Chr.9: 35657681-35657681 on Build GRCh38
rs756285919 Chr.9: 35657869-35657869 on Build GRCh38
rs778202312 Chr.9: 35657771-35657771 on Build GRCh38
rs544206655 Chr.9: 35657592-35657592 on Build GRCh38
rs758130879 Chr.9: 35657895-35657895 on Build GRCh38
rs982511437 Chr.9: 35657582-35657582 on Build GRCh38
rs773432147 Chr.9: 35657822-35657822 on Build GRCh38
rs769080777 Chr.9: 35657758-35657758 on Build GRCh38
rs1053360711 Chr.9: 35657960-35657961 on Build GRCh38
rs145720742 Chr.9: 35657806-35657806 on Build GRCh38
rs377566038 Chr.9: 35657843-35657843 on Build GRCh38
rs753874439 Chr.9: 35657872-35657872 on Build GRCh38
rs940193083 Chr.9: 35657927-35657927 on Build GRCh38
rs928825069 Chr.9: 35657881-35657881 on Build GRCh38
rs542778133 Chr.9: 35657913-35657913 on Build GRCh38
rs193117694 Chr.9: 35657965-35657965 on Build GRCh38
rs1051076135 Chr.9: 35657833-35657833 on Build GRCh38
rs539893970 Chr.9: 35657668-35657668 on Build GRCh38
rs387906533 Chr.9: 35657927-35657928 on Build GRCh38
rs113967059 Chr.9: 35657876-35657876 on Build GRCh38
rs954709065 Chr.9: 35657917-35657917 on Build GRCh38
rs902342946 Chr.9: 35657961-35657961 on Build GRCh38
rs773388104 Chr.9: 35657731-35657731 on Build GRCh38
rs573644542 Chr.9: 35657901-35657901 on Build GRCh38
rs535595484 Chr.9: 35657860-35657860 on Build GRCh38
rs964504772 Chr.9: 35657816-35657816 on Build GRCh38
rs772665785 Chr.9: 35657805-35657805 on Build GRCh38
rs312262896 Chr.9: 35657948-35657949 on Build GRCh38
rs183570217 Chr.9: 35657817-35657817 on Build GRCh38
rs752709977 Chr.9: 35657864-35657864 on Build GRCh38
rs889473053 Chr.9: 35657942-35657942 on Build GRCh38
rs932549258 Chr.9: 35657661-35657661 on Build GRCh38
rs1043977324 Chr.9: 35657619-35657619 on Build GRCh38
rs747605028 Chr.9: 35657904-35657904 on Build GRCh38
rs866678717 Chr.9: 35657712-35657712 on Build GRCh38
rs774079455 Chr.9: 35657914-35657914 on Build GRCh38
rs1023945900 Chr.9: 35657563-35657563 on Build GRCh38
rs182717768 Chr.9: 35657572-35657572 on Build GRCh38
rs565056797 Chr.9: 35657846-35657846 on Build GRCh38
rs527471640 Chr.9: 35657820-35657820 on Build GRCh38
rs572893111 Chr.9: 35657932-35657932 on Build GRCh38
rs983337094 Chr.9: 35657918-35657918 on Build GRCh38
rs557906740 Chr.9: 35657753-35657753 on Build GRCh38
rs530627656 Chr.9: 35657852-35657852 on Build GRCh38
rs781079047 Chr.9: 35657766-35657766 on Build GRCh38
rs746248517 Chr.9: 35657880-35657880 on Build GRCh38
rs963319834 Chr.9: 35657922-35657922 on Build GRCh38
rs549849356 Chr.9: 35657899-35657899 on Build GRCh38
rs745416646 Chr.9: 35657937-35657937 on Build GRCh38
rs775287390 Chr.9: 35657912-35657913 on Build GRCh38
rs928762713 Chr.9: 35657926-35657926 on Build GRCh38
rs151011853 Chr.9: 35657962-35657963 on Build GRCh38
rs187324737 Chr.9: 35657779-35657779 on Build GRCh38
rs752723025 Chr.9: 35657928-35657928 on Build GRCh38
rs551655682 Chr.9: 35657749-35657749 on Build GRCh38
rs757576534 Chr.9: 35657873-35657873 on Build GRCh38
rs796065036 Chr.9: 35657823-35657824 on Build GRCh38
rs763923256 Chr.9: 35657902-35657902 on Build GRCh38
rs749839597 Chr.9: 35657746-35657746 on Build GRCh38
rs570362765 Chr.9: 35657581-35657582 on Build GRCh38
rs748268184 Chr.9: 35657767-35657767 on Build GRCh38
rs774897161 Chr.9: 35657787-35657787 on Build GRCh38
rs903310510 Chr.9: 35657707-35657707 on Build GRCh38
rs557578605 Chr.9: 35657898-35657898 on Build GRCh38
rs751373603 Chr.9: 35657858-35657858 on Build GRCh38
rs962530896 Chr.9: 35657595-35657595 on Build GRCh38
rs915919453 Chr.9: 35657921-35657921 on Build GRCh38
rs1004469515 Chr.9: 35657838-35657838 on Build GRCh38
rs548292581 Chr.9: 35657633-35657633 on Build GRCh38
rs886041234 Chr.9: 35657804-35657804 on Build GRCh38
rs750141239 Chr.9: 35657842-35657842 on Build GRCh38
rs188907984 Chr.9: 35657839-35657839 on Build GRCh38
rs767355005 Chr.9: 35657925-35657925 on Build GRCh38
rs757755234 Chr.9: 35657745-35657746 on Build GRCh38
rs908555769 Chr.9: 35657834-35657834 on Build GRCh38
rs768594644 Chr.9: 35657935-35657936 on Build GRCh38
rs1040824418 Chr.9: 35657960-35657960 on Build GRCh38
rs370797103 Chr.9: 35657857-35657857 on Build GRCh38
rs765138079 Chr.9: 35657884-35657884 on Build GRCh38
rs578091934 Chr.9: 35657879-35657879 on Build GRCh38
rs962412737 Chr.9: 35657754-35657754 on Build GRCh38
rs1020582110 Chr.9: 35657849-35657849 on Build GRCh38
rs1021056174 Chr.9: 35657801-35657801 on Build GRCh38
rs770577795 Chr.9: 35657677-35657677 on Build GRCh38
rs1039657685 Chr.9: 35657666-35657666 on Build GRCh38
rs907490025 Chr.9: 35657829-35657829 on Build GRCh38
rs549047941 Chr.9: 35657746-35657746 on Build GRCh38
rs903769824 Chr.9: 35657638-35657638 on Build GRCh38
rs957362192 Chr.9: 35657916-35657916 on Build GRCh38
rs7021463 Chr.9: 35657862-35657862 on Build GRCh38
rs940080676 Chr.9: 35657838-35657839 on Build GRCh38
rs149105942 Chr.9: 35657863-35657863 on Build GRCh38
rs1031995271 Chr.9: 35657908-35657908 on Build GRCh38
rs1032219537 Chr.9: 35657964-35657964 on Build GRCh38
rs1051465260 Chr.9: 35657664-35657664 on Build GRCh38
rs948931144 Chr.9: 35657823-35657823 on Build GRCh38
rs377349293 Chr.9: 35657748-35657748 on Build GRCh38
rs759632477 Chr.9: 35657874-35657874 on Build GRCh38
rs1002409010 Chr.9: 35657784-35657784 on Build GRCh38
rs549256667 Chr.9: 35657549-35657549 on Build GRCh38
rs571254176 Chr.9: 35657900-35657900 on Build GRCh38
rs727502774 Chr.9: 35657756-35657756 on Build GRCh38
rs535022008 Chr.9: 35657752-35657752 on Build GRCh38
rs537096111 Chr.9: 35657592-35657593 on Build GRCh38
rs779796897 Chr.9: 35657819-35657819 on Build GRCh38
rs758198618 Chr.9: 35657616-35657616 on Build GRCh38
rs769331218 Chr.9: 35657905-35657905 on Build GRCh38
rs1000522852 Chr.9: 35657861-35657861 on Build GRCh38
rs977992022 Chr.9: 35657870-35657870 on Build GRCh38
rs890235700 Chr.9: 35657970-35657970 on Build GRCh38
rs199476103 Chr.9: 35657948-35657948 on Build GRCh38
rs560328768 Chr.9: 35657791-35657791 on Build GRCh38
rs970510851 Chr.9: 35657923-35657923 on Build GRCh38
rs748911449 Chr.9: 35657921-35657922 on Build GRCh38
rs7021642 Chr.9: 35657841-35657841 on Build GRCh38
rs569993924 Chr.9: 35657704-35657704 on Build GRCh38
rs962926307 Chr.9: 35657716-35657716 on Build GRCh38
rs949582348 Chr.9: 35657940-35657940 on Build GRCh38
rs1042583858 Chr.9: 35657941-35657941 on Build GRCh38
rs545284014 Chr.9: 35657933-35657933 on Build GRCh38
rs578123788 Chr.9: 35657755-35657755 on Build GRCh38
rs771464779 Chr.9: 35657782-35657782 on Build GRCh38
rs912498335 Chr.9: 35657777-35657777 on Build GRCh38
rs761398394 Chr.9: 35657825-35657825 on Build GRCh38
rs767056929 Chr.9: 35657770-35657771 on Build GRCh38
rs1013594536 Chr.9: 35657537-35657537 on Build GRCh38
rs1049483652 Chr.9: 35657892-35657892 on Build GRCh38
rs999349961 Chr.9: 35657963-35657963 on Build GRCh38
rs1029736982 Chr.9: 35657697-35657697 on Build GRCh38
rs762145032 Chr.9: 35657788-35657788 on Build GRCh38
rs780957606 Chr.9: 35657930-35657930 on Build GRCh38
rs370303129 Chr.9: 35657813-35657813 on Build GRCh38
rs565037825 Chr.9: 35657938-35657938 on Build GRCh38
rs192060920 Chr.9: 35657807-35657807 on Build GRCh38
rs945227726 Chr.9: 35657612-35657612 on Build GRCh38
rs1030486815 Chr.9: 35657969-35657969 on Build GRCh38
rs530883024 Chr.9: 35657953-35657953 on Build GRCh38
rs536333075 Chr.9: 35657919-35657919 on Build GRCh38
rs925085494 Chr.9: 35657814-35657814 on Build GRCh38
rs1013239356 Chr.9: 35657798-35657798 on Build GRCh38
rs567665484 Chr.9: 35657910-35657910 on Build GRCh38
rs913676224 Chr.9: 35657601-35657601 on Build GRCh38
rs112804667 Chr.9: 35657575-35657575 on Build GRCh38
rs898143345 Chr.9: 35657887-35657887 on Build GRCh38
rs902757638 Chr.9: 35657943-35657943 on Build GRCh38
rs763426582 Chr.9: 35657740-35657740 on Build GRCh38
rs1038523315 Chr.9: 35657778-35657778 on Build GRCh38
rs995230090 Chr.9: 35657848-35657848 on Build GRCh38
rs924622406 Chr.9: 35657797-35657797 on Build GRCh38
rs1045976155 Chr.9: 35657826-35657826 on Build GRCh38
rs534955648 Chr.9: 35657868-35657868 on Build GRCh38
rs773125557 Chr.9: 35657930-35657930 on Build GRCh38
rs936059863 Chr.9: 35657800-35657800 on Build GRCh38
rs74810894 Chr.9: 35657891-35657891 on Build GRCh38
rs769131655 Chr.9: 35657904-35657905 on Build GRCh38
rs745372450 Chr.9: 35657866-35657867 on Build GRCh38
rs786204684 Chr.9: 35657955-35657955 on Build GRCh38

View Full Product Details