Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004286.4 | NP_004277.2 | 1 - 2 | 350 | 85 |
XM_011530537.2 | XP_011528839.1 | 1 - 2 | 1588 | 85 | |
XM_017029099.1 | XP_016884588.1 | 1 - 2 | 1580 | 85 | |
XM_017029100.1 | XP_016884589.1 | 1 - 2 | 1574 | 85 | |
XM_017029101.1 | XP_016884590.1 | 1 - 2 | 94 | 85 | |
GenBank mRNA | BC014075.1 | - | 1 - 2 | 188 | 85 |
CR456501.1 | - | 1 - 2 | 173 | 85 | |
U87964.1 | - | 1 - 2 | 94 | 85 |