Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47215176-47215305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130959 c.111C>G p.L37L Chr.11: 47215247-47215247 on Build GRCh38
5895260 c.53_54insC p.R20fs*26 Chr.11: 47215189-47215190 on Build GRCh38
1981286 c.43A>C p.I15L Chr.11: 47215179-47215179 on Build GRCh38
6581410 c.126C>G p.S42S Chr.11: 47215262-47215262 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775814662 Chr.11: 47215239-47215239 on Build GRCh38
rs530733334 Chr.11: 47215240-47215240 on Build GRCh38
rs754405766 Chr.11: 47215199-47215199 on Build GRCh38
rs148175447 Chr.11: 47215215-47215215 on Build GRCh38
rs770855579 Chr.11: 47215233-47215233 on Build GRCh38
rs776815495 Chr.11: 47215258-47215258 on Build GRCh38
rs777698820 Chr.11: 47215217-47215217 on Build GRCh38
rs368073045 Chr.11: 47215248-47215248 on Build GRCh38
rs772542105 Chr.11: 47215257-47215257 on Build GRCh38
rs956219237 Chr.11: 47215205-47215205 on Build GRCh38
rs780776638 Chr.11: 47215213-47215213 on Build GRCh38
rs199965459 Chr.11: 47215268-47215268 on Build GRCh38
rs749034260 Chr.11: 47215219-47215219 on Build GRCh38

View Full Product Details